Thermo Fisher Scientific
OMNIC Advanced Mapping Options Not Showing
Selecting Advanced Mapping Options in OMNIC/OMNIC for Dispersive Raman may show nothing if Window Styles 2 or 4 are active; switch to styles 1, 3, or 5 to display Start and End Position fields for line/area maps (not for point maps).
Issue
When the advanced mapping options check box is selected on the Experiment Setup - Mapping tab, the additional boxes are not displayed
Environment
OMNIC
OMNIC For Dispersive Raman
Resolution
Window Styles 2 and 4 in OMNIC do not allow for the advanced mapping boxes to be displayed.  To change the Window Style:
  1. Click on the Edit menu and Options command
  2. Select the View tab
  3. Click the Window Style button to change the Window style.
  4. Click Ok
  5. Check if Advanced Mapping Options now display.
  6. To make the change permanent, click the File menu and Save Configuration As to save the configuration file.
Edit Options - View - Window Style.jpg
Analysis
On the Mapping tab of the Experiment setup tab, there is a check box for displaying extra boxes to line or area maps.  Note that there no additional boxes for point maps.
OMNIC Experiment Setup - Advanced Mapping Options Unchecked.jpg
When the Advanced mapping options box is checked, additional boxes for "Start Position" and "End Position" should be displayed under the Dimension section at the top of the window.
OMNIC Experiment Setup - Advanced Mapping Options Checked.jpg
There are 5 possible Window Styles available in the OMNIC software.  Styles 1, 3, and 5 allow for the Advanced Mapping Options to be displayed, while styles 2 and 4 do not.  The images below show the 5 different window styles available in OMNIC.
Style 1 OMNIC Windows Style 1.jpg
Style 2 OMNIC Window Style 2.jpg
Style 3 OMNIC Window Style 3.jpg
Style 4 OMNIC Window Style 4.jpg
Style 5 OMNIC Window Style 5.jpg
  • Was this article helpful?