Output Data to a Text File from TQ Analyst Software
Issue
How do you output data to a text file?
Environment
TQ Analyst
Resolution
TQ Analyst can output data to a text file:
Prepending answers to a text file will add a new line at the top of the file with the answer. The information can then be accessed by a LIMS or other network archiving service.
The file and path that TQ prepends data to: C:\Users\”active user”\AppData\Local\Temp\TQInputZ.txt
Attachment(s)
| File | Last Modified |
|---|---|
| Prepend_answers_to_text_file.png | March 11, 2022 |