Uninstalling OMNIC Paradigm
Description:
To uninstall OMNIC Paradigm, ThermoMSIUninstall should not be used if the uninstall version is prior to version 4.10.
Omnic Paradigm software starting with v1.2HF1 includes the first version of ThermoMSIUninstall that will work with Paradigm which is Version 4.10; before Paradigm v1.2HF1, uninstall was not a part of Paradigm software.
When installing an upgrade, the previous version of Paradigm does not need to be removed. Upgrades can just install over the existing version.
Environment:
OMNIC Paradigm