Thermo Fisher Scientific
Clean the sample windows
Ensure sample windows are clean before and after using the Affirma Lubricant Analysis system. Required materials include heptane or isooctane, a clean dropper bottle, non-abrasive wipes, and nitrile gloves. Avoid chemical exposure by wearing gloves and disposing of soiled materials properly. Clean windows gently to avoid damage.
Ensure sample windows are clean before and after using the Affirma Lubricant Analysis system. Required materials include heptane or isooctane, a clean dropper bottle, non-abrasive wipes, and nitrile gloves. Avoid chemical exposure by wearing gloves and disposing of soiled materials properly. Clean windows gently to avoid damage.
Make sure the sample windows are clean before and after taking sample measurements with the Affirma Lubricant Analysis system.
Materials required:
  • Small volume of heptane or isooctane
  • Clean dropper bottle for cleaning solvent (provided)
  • Clean, non-abrasive laboratory wipes (unscented and lotion-free; paper napkins work well)
  • Powder-free nitrile gloves
caution-generic.png
Avoid chemical exposure. Isooctane and heptane are chemical irritants. When handling isooctane or heptane: 
  • do not ingest
  • avoid contact with skin and eyes
  • avoid inhaling vapor or mist
  • wear gloves that provide adequate protection
  • properly dispose of soiled containers, wipes, gloves, etc.
When cleaning the sample windows, avoid using excessive force or you may damage the crystal.
To clean the sample windows
  1. Open the sample tray, raise the top window housing a small amount, place a clean, non-abrasive laboratory wipe between the two windows and then lower the housing to soak up most of the material.
  2. Move the top window housing to the load position and use a new wipe to gently clean the upper and lower sample windows and surrounding areas.
  3. Place a few drops of cleaning solvent onto a new wipe and use it to thoroughly clean both windows and the area around them.
  4. To protect the sampling surfaces, place the top window housing in the sampling position and close the sample tray.
  • Was this article helpful?