Thermo Fisher Scientific
Install the software and Affirma workflow package
To install the Affirma workflow package, run the Setup.exe file for your accessory and reboot your computer. Start OMNIC Paradigm software, connect to your instrument, and switch to Operator mode. Enter Administrator mode, navigate to the Factory Packages folder, and open the .rdep file for your accessory. Exit Operator mode to access the Affirma workflows.
To install the Affirma workflow package, run the Setup.exe file for your accessory and reboot your computer. Start OMNIC Paradigm software, connect to your instrument, and switch to Operator mode. Enter Administrator mode, navigate to the Factory Packages folder, and open the .rdep file for your accessory. Exit Operator mode to access the Affirma workflows.
Follow the steps below to install the Affirma workflow packages.
To install the Affirma workflow package
  1. Run the Setup.exe file to install the corresponding Affirma package for your accessory, then reboot your computer. Each accessory has its own file.
  2. Start OMNIC Paradigm software.
  3. Connect to your instrument. To do so, go to Configure > Connectivity > Nicolet Summit.
  4. Go to Configure > View > Operator. This will put the software in Operator mode.
  5. In the upper left-hand corner, select the triple bar button, then choose Administrator and enter the password.
  6. Select Open in the upper right-hand side of the screen and select the following path:
C:\Users\Public\ Documents\Thermo Scientific\OMNIC Paradigm\Factory Packages
Figure 1-1: Open folder icon in OMNIC Paradigm software Operator mode
affirma1.png
Figure 1-2: OMNIC Paradigm software Operator mode Factory Packages location
affirma3.png
  1. Select the .rdep file for your accessory and select Open.
The .rdep file will only appear if you have followed these steps in order and installed the appropriate Setup.exe file.
  1. In the upper left-hand corner, select the triple bar button, then select Home to exit Operator mode. The Affirma workflows for your accessory are now available in Operator mode.
Figure 1-3: OMNIC Paradigm software home screen with Affirma workflows
affirma4.png
  • Was this article helpful?