Thermo Fisher Scientific
Importing Spectral Data from Files
Once you have uploaded spectral files to Data Connect, import their data to OMNIC Anywhere by navigating to the Measurements page, clicking on Import measurement, selecting Data Connect files, choosing a folder with spectral files, adding the desired files, and clicking Import to add the data to OMNIC Anywhere.
Once you have uploaded spectral files to Data Connect, import their data to OMNIC Anywhere following these steps:
  1. In the Measurements page, click on Import measurement
  2. Click on Data Connect files
    OMNIC Anywhere import data 1.png
  3. Select a folder that includes spectral files
  4. Click + Add on any files you want to import
    OMNIC Anywhere import data 2.png
  5. Click Import to add your spectral data to OMNIC Anywhere
  • Was this article helpful?