Performing a Correlation Search
Double-click on any measurement in the Measurements page and click on Correlation search. Select your libraries for the search and click Next. Optionally, select a region for the search and set your search parameters. Finally, click Run search to perform the correlation search.
- Double-click on any measurement in the Measurements page
-
Click on Correlation search
- Select your libraries for search and click Next
- Select a region for search and click Next (optional)
- Set your search parameters (optional)
-
Click Run search