Thermo Fisher Scientific
Uploading Spectral Files
To import spectral data to OMNIC Anywhere, go to the Measurements screen and click Import measurement. Drag and drop your spectral files or click Upload files to select them manually. Confirm the upload, select the storage location, and click Upload. The Status column indicates sync status: blue for synced, gray for unsynced, and red for errors. To resync, click the red cloud icon and Sync Again.
To import your spectral data to OMNIC Anywhere, follow these steps:
1. In the Measurements screen, click on Import measurement.
Screenshot 2024-05-29 at 1.09.26 PM.png
2. Drag and drop your spectral files to the assigned area or click on Upload files to select manually.
Screenshot 2024-05-29 at 1.09.54 PM.png
3. Click on Confirm.
Screenshot 2024-05-29 at 1.10.19 PM.png
4. Select the location the location where you want to store your files. This will backup your files in your cloud Data Storage.
5. Click on Upload.
Screenshot 2024-05-29 at 1.10.37 PM.png
6. A notification will be displayed and measurements will appear in your Measurements table when your spectral files have been uploaded and are ready to use in OMNIC Anywhere.
Screenshot 2024-05-29 at 1.13.47 PM.png
Status column
The Status column lets you easily know the sync status of your measurement:
  • A blue icon represents a synced status, meaning your original file is stored in Data Storage and your Measurement can be used correctly.
  • A gray icon represents an unsynced status, meaning your original file is not stored in Data Storage, but your Measurement can still be used.
  • A red icon represents a syncing error, meaning there was an issue when syncing your file to OMNIC Anywhere. Your original file will still be backed up in the cloud, but you will not be able to use your measurement until the sync issue is corrected.
Screenshot 2024-05-30 at 11.05.51 AM.png
To resync an unsynced file, click on the red cloud icon, then click on Sync Again.
Screenshot 2024-05-30 at 11.32.54 AM.png
  • Was this article helpful?