Measurements Not Being Displayed on Dashboard and "Site Can't Be Reached" Error When Running Workflows
OMNIC Paradigm software may experience three issues: delayed opening, missing new background/sample data, and workflows returning a "This site can't be reached" error. These problems can be resolved by turning off the Memory Integrity setting in Windows 11. To do this, access "Core Isolation" via the Windows Settings App, disable the Memory Integrity toggle in the Windows Security app and reboot the computer.
Issue
There are three separate issues that may affect OMNIC Paradigm software that can be caused by the same setting in Windows.
- Paradigm takes longer than normal to open.
- After collecting a new background or sample, it doesn't show up in the list of measurements and/or previously collected spectra are not shown.
- Running a workflow gives an error "This site can't be reached".
Environment
OMNIC Paradigm
Resolution
Turn off the Memory Integrity setting in Windows 11 and reboot the computer.
- Click on the Windows Start menu and enter "Core Isolation".
- Click on "Core Isolation" to open Windows Security app.
- On the Core Isolation page, turn OFF the toggle for Memory Integrity.
- Reboot the PC.
Analysis
To nagivate to this setting manually, select Start > Settings > Update & Security > Windows Security > Device Security and then under Core isolation, selecting Core isolation details.
Attachment(s)
| File | Last Modified |
|---|---|
| Security - Core Isolation Settings.jpg | October 19, 2025 |
| site cannot be reached.jpg | October 19, 2025 |
| no measurements.jpg | October 19, 2025 |