Paradigm Peak Height, Peak Area Tools
In OMNIC Paradigm, peak area and peak height tools are accessed by right-clicking in spectral view and selecting them from the Display Tools submenu; results are saved in the spectrum history.
Issue
OMNIC Paradigm does not have palete tools like OMNIC for Peak Area or Peak Height and the functions are not listed n the menus.
Environment
OMNIC Paradigm
Resolution
The locate the functions, while in spectral view, right click in the window to get a sub menu. Expand display tools to see the functions.
After performing the function and saving the spectra, the results will be in the history of the spectra.
Attachment(s)
| File | Last Modified |
|---|---|
| Paradigm peak area tools.png | December 22, 2021 |