Steps to change or update standards or expiration date in OMNIC System status
This article provides the steps to change or update standards or expiration date in OMNIC System status.
Issue
Steps to change or update standards or expiration date in Omnic System status
Environment
- Omnic Software
Resolution
Here are the steps to change the standard expiration date:
- Open the Omnic Software:
- Launch the Omnic software on your computer.
- Access the System Status:
- Navigate to the 'View' menu and select 'Configure System Status'.
- Locate Valpro Qualification or Performance verification:
- Then click on the Configure Standards
- Select the Standard or click add for a new standard
- Find and select the standard for which you want to change the expiration date.
- Edit Standard Information:
- There should be an option to update the standard information. Click on this option.
- Change Expiration Date:
- Locate the field for the expiration date. Enter the new expiration date as required.
- Save Changes:
- Save the changes you have made by click ok. Ensure that the new expiration date is correctly displayed.
- Verify:
- Verify that the new expiration date has been updated and is reflected correctly in the system.