Thermo Fisher Scientific
Renaming spectral libraries in OMNIC
Description: 
OMNIC does not have a direct way to change the name of a Search or QC library. The workaround is to create a new library with the same set of custom information fields and spectral range as the original library. The attached document describes the process for creating the new library.
Environment:
OMNIC
  • Was this article helpful?