Measure a microscopy sample
Use OMNIC™ Paradigm™ software to operate the microscope and to analyze samples. This section describes a basic measurement process using Reflection or Transmission analysis.
For detailed information on measurement settings and software controls, refer to the OMNIC™ Paradigm™ software user manual.
Before you begin
Verify the following:
-
The sample is prepared.
-
The software is running.
-
The instrument is ready
Load the sample
Eject the stage so that you have more access to the stage to position your sample. If your sample is small and fits in position easily, you can position the sample without ejecting the stage.
-
In the software, click Eject Stage. Ejecting the stage lowers the stage to make it easier for you to load a sample. If you have a manual stage, lower it using the joystick or software controls.
-
Insert the sample slide. The stage fits a universal sample holder.
Figure 53. Sample insert
View the sample
Click Start to open a new Microscope tab and view your sample. In the Microscope tab, the camera view shows a live image of the sample. Use the camera tools or the optional joystick to adjust the illumination and bring the sample into focus.
Locate and light the sample
Use the camera view or optional joystick to adjust illumination and bring the sample into focus. For detailed instructions on illumination and focus controls, refer to the OMNIC™ Paradigm™ Software user manual.
-
Open the camera view and open the Focus settings tab.
Camera view focus settings
-
Select Autofocus. The stage moves up and down to find the optimal focus. Autofocus works best with areas of high visual contrast. Autofocus may struggle with some low-contrast samples and samples with multiple focal planes. Autofocus also automatically lights the sample.
Optional: capture a mosaic
A mosaic is a collection of pictures of the sample that are stitched together into a single large image. The mosaic serves as the main workspace for navigating the sample and identifying areas of interest.
Systems with a manual stage cannot capture a mosaic. Use the camera view to navigate the sample.
Before you capture a mosaic, make sure that the sample is in focus and illuminated.
-
Select the Draw Mosaic tool from toolbar.
-
Click and drag on the mosaic pane to draw a mosaic region.
-
Select Capture Mosaic.
Measure the background and sample
Review your measurement and background settings in the left panel.
|
Select Live Spectrum to continuously view the single beam spectrum as move around the sample. This can help you determine if you want to use a particular point for your background measurement. If you select this option, the software displays a live single beam spectrum in the spectra pane. |
-
Move the stage to the reference point for your analysis type.
-
Select Background from the toolbar. When the measurement is complete, the background spectrum is stored in the database and added to the spectra pane.
If you have a mosaic, select a point on the mosaic to automatically move the stage to that location.
If you do not have a mosaic, use the camera view or joystick to navigate to the point of interest.
-
Switch to the 15x objective. To change objectives, select Change Objective in the toolbar, in the camera view, select the Objectives tab.
-
Select Sample from the toolbar.
When the measurement is complete, the spectrum is stored in the database and added to the spectra pane. You can now process, identify, and analyze the spectrum. For more information about OMNIC™ Paradigm™ software’s processing and search tools, refer to the OMNIC™ Paradigm™ software user manual.