Thermo Fisher Scientific
Measure with ATR
With the optional attenuated total reflection (ATR) attachment, you can analyze highly infrared-absorbant or hard-to-prepare microscopic materials, often with little or no sample preparation. Examples of these materials include polymers, coatings, rubbers, coated papers, and biological materials.
ATR measurements are not compatible with particle analysis in the software. If you are performing particle analysis, use reflection or transmission techniques instead.
NOTICE
The ATR tip may leave a small indentation on the sample surface. This is normal and does not indicate damage to the ATR crystal.
Applications of ATR microscopy include:
  • Analyzing the surface of a sample.
  • Analyzing highly-absorbing materials and the surfaces of thick samples.
  • Analyzing surface coatings.
  • Analyzing surface defects, inclusions, or degradation.
Installing the ATR attachment
The ATR attachment is held to the objective using magnets. To install the attachment, align the indicator dots on the objective with the dots on the attachment. The recessed spaces on the attachment must line up with the guides on the objective.
atr-mount2.jpg
Table 6. The ATR attachment
Label Feature Description
A
Alignment indicators on the objective
The objective has three indicators: A single dot, two dots, and three dots. These help you align the attachment correctly.
B
Alignment indicators on the ATR attachment
Using these alignment indicators on the attachment, match the single dot on the objective with the single dot on the attachment, the two dots with the two dots, and the three dots with the three dots. This will ensure that the ATR is properly aligned for use.
C
QR codes
The ATR sensor reads QR codes on the ATR attachment to automatically recognize which ATR is installed and to recognize the position of the attachment.
Measure data using ATR
Measuring data with the ATR attachment follows the same processes as other types of measurements. When you set "Analyze using" to ATR, you are prompted to insert the ATR crystal attachment as needed.
 To measure using ATR
  1. On the dashboard, select ATR from the Analyze using list.
    Analyze using ATR
    atr-analyze-using.png
  2. Optional: Review and edit the ATR Contact settings.
    1. In the Camera view, select the ATR tab to view the ATR settings.
      ATR contact
      atr-settings.png
    Setting Description
    Target pressure
    This is the target pressure that will be applied during measurement. Click and drag the slider or enter an exact value.
    Release distance
    The vertical distance that the stage moves once the ATR contact is released. A larger distance provides more clearance but will cause the ATR measurement to take longer, since the stage will move farther at every point.
    Contact
    Press to test contact.
    Release
    Press to release contact.
    Profile
    Select the settings use to use for the ATR contact and release.
  3. Measure a background and your sample as normal. Target pressure is automatically applied during the measurement. You are prompted to install the attachment when needed.
The ATR attachment can be used for individual measurements or for measuring regions in a mapping session.
ATR clean check
Run a manual "clean check" in OMNIC™ Paradigm™ software to check the cleanliness of an ATR crystal after completion of an experiment. A comparison is made between a clean sample and a contaminated one, which is captured via peaks in the spectra. As this is a manual process, the ATR clean check involves certain necessary steps that would not be normally required. An eggshell business card would be an adequate sample for this type of check.
The process involves collecting a spectrum of your sample, then releasing the pressure (by raising the crystal off of the sample) and measuring another spectrum. If the crystal has picked up contamination from the sample, such as oil or adhesive residue, the spectrum of the crystal will show residual peaks. This then requires cleaning the crystal.
 To run an ATR clean check
  1. Place your sample and set your parameters.
    1. "Analyze using" must be set to ATR.
    2. Set measure background to "After time." Both sample measurements in this procedure should use the same background measurement.
  2. In a new microscope tab, open the camera view.
  3. Select Change Objective to change to the IR capable 15x objective.
  4. In the camera view, select the stage tools tab and choose Show Crosshairs to aid in seeing your target.
      A message appears at this point if the ATR crystal attachment is not installed. If this occurs, simply insert the attachment and select OK (the option will be unavailable until the attachment is inserted).
    ATR attachment pop-up message
    atr-insert.png
  5. Under the ATR tab, set the target pressure and release distance. The suggested settings for the purposes of the check are a target pressure of 50 and a release distance of 3500 microns, but you are free to set different values if preferred. A large release distance ensures that the crystal comes clear of the sample.
  6. Measure a background spectrum. You can do this using the Background tool in the toolbar. Measure the background as normal. The ATR crystal should not be in contact with the sample while collecting the background.
  7. When the background is complete, select Contact under the ATR Contact menu to manually bring the sample into contact with the ATR. This does not move the stage.
  8. Select Sample to measure the spectrum.
  9. Select Release under the ATR Contact menu to release the crystal, then select Measure again to obtain a measurement of the crystal itself.
    Examine the spectrum from the released crystal. If there are peaks present, it indicates contamination from the sample. Shown here is a residual oil. If the spectrum is essentially flat, you can continue. Otherwise, you should clean the crystal before proceeding.
    Residual oil sample
    ATR-clean-check.png
  • Was this article helpful?