Navigate workflows
This article provides an overview of these basic workflow features and tools
-
Workflow pane
-
Run Workflow window
-
Workflow editor
For more information on when and why to use workflows, refer to "Introduction to workflows."
Workflow pane
The workflow pane is always available at the bottom of the OMNIC Paradigm dashboard. Use this space to preview and run workflows you’ve created, and to open the workflow editor to edit or create a new workflow.
| Label | Area | Description |
|---|---|---|
|
A |
Dashboard tabs |
Select the Workflows tab to view workflows and packages. |
|
B |
Data list |
Select Workflows from the list. |
|
C |
Context menu |
Right-click a workflow to view more options and commands. |
|
D |
Workflow preview |
This area displays a preview of the workflow. If the workflow is locked, this shows the workflow logo instead of the preview. |
| To do this… | Take this action |
|---|---|
|
Select and preview a workflow |
Click once on the workflow name |
|
Test or run a workflow |
Right-click the workflow and select Run. This opens the Run Workflow window where you can step through the workflow tasks. |
|
Edit, rename, duplicate, delete or export the selected workflow |
Right-click the workflow and choose an option from the shortcut menu. |
Run workflow window
Use the Run Workflow window to test or run a workflow. To open the run Workflow window, right-click a workflow and select Run.
From the Run Workflow window, use the arrow button to move to the next task in the workflow. The workflow will run exactly as it has been designed, with tasks and operator messages displayed as indicated by the workflow.
To stop the workflow and return to the dashboard, click the Stop button in the Play Workflow window. (If the workflow does not have a Stop button, click the Dashboard button on the toolbar to stop the workflow.)
Workflow editor
The workflow editor contains all the tools for creating workflows, including the workflow menu, toolbar, tile selection bar, and canvas. To open the workflow editor, right-click a workflow name on the dashboard and choose Edit from the shortcut menu.
You can also open the workflow editor from the OMNIC Paradigm main window by choosing:
File > Workflow > Create
The workflow canvas is where you select and set up the tasks you want the workflow to perform and join tasks together to create the workflow. The workflow editor opens with the mandatory “Workflow Start” and “Workflow End” tiles displayed on the canvas.
| Label | Area | Description |
|---|---|---|
|
A |
Toolbar |
Frequently used tools. |
|
B |
Tiles |
Drag and drop tiles to the canvas to add them to your workflow. |
|
C |
Workflow canvas |
This is where you build your workflow. |
|
D |
Target instrument list |
Select an instrument from the list to associate it with this workflow. The workflow will run only on instruments of the target type. |
For details on creating and editing workflows, refer to "Create and run your first workflow."