Simulate an instrument
The OMNIC Paradigm software includes a built-in instrument simulator that allows you to use the software without being connected to a physical instrument. The simulator is useful for training new users on the software, developing workflows, and experimenting with different instrument settings without needing to have a spectrometer connected.
-
From the dashboard, go to the Configure menu and select Connectivity.
-
In the Select Instrument Connection dialog, select Simulator and click Connect.
-
Select an instrument to simulate from the list of available instruments and click Ok.
The simulator runs as if you were connected to a real instrument. You can acquire data, run workflows, and experiment with different instrument settings. Note that not all instrument features are available in the simulator. For example, you cannot simulate some diagnostic tests.
When the simulator is running, the instrument status icon in the upper right corner of the software displays the text "Simulation" after the instrument type.