OMNIC Paradigm Software Tiers
The OMNIC Paradigm software is available in three tiers.
|
|
Tier 1 |
Tier 2 |
Tier 3 |
Description |
|
Desktop & Touchscreen views |
Basic Spectroscopy |
Basic Spectroscopy |
Basic Spectroscopy |
Includes basic tools for collecting, analyzing, and processing spectra. |
|
Operator view |
Custom Solutions |
Custom Solutions |
Custom Solutions |
Develop custom solutions with the package editor and Operator view. Packages include workflows and their required assets, and the Operator view provides a streamlined interface for users to run only your assigned workflows. |
|
Desktop view only |
- |
Time Series Measurements |
Time Series Measurements |
Use Time Series measurements to measure your sample as it changes over time, such as in a GCIR or TGA-IR experiment. |
|
- |
- |
FTIR Microscopy Sessions |
Use microscopy session tools to view a sample surface and to measure regions, lines, points, and particles. |
|
|
- |
- |
FT-Raman Tools |
The FT-Raman software tools allow you to measure data with the FT-Raman module. |
|
|
|
TQ Analyst EZ Software |
TQ Analyst PRO Software |
TQ Analyst PRO Software |
EZ Edition: This software is a sophisticated yet easy to use software package for developing analytical methods for spectral applications, including mid-infrared, near-infrared, far-infrared, and Raman.
PRO Edition: This standalone software is a comprehensive method development platform with all the performance and flexibility to develop robust methods. It offers a complete set of tools for spectral measurement, classification and quantitative analysis. |
|
Computer Requirements |
||||
|
|
Intel Core i5 (6 cores, 3.3GHz to 4.8GHz) |
Intel Core i7 (8 cores, 2.9GHz to 4.8GHz) |
Intel Core i7 (8 cores, 2.9GHz to 4.8GHz) |
Processor |
|
Minimum: 8 GB Recommended: 16 GB |
16 GB |
32 GB |
RAM |
|
|
Standard integrated graphics card |
Validated graphics card[1] |
Validated graphics card |
Graphics Card |
|
|
500 GB HDD |
1 TB HDD |
1 TB SDD |
Hard Disk Space |
|
|
Mouse, Keyboard, Monitor |
Mouse, Keyboard, Monitor |
Mouse, Keyboard, Monitor |
Peripherals |
|
|
A DVD drive is required for software installation for all computer options |
Disk Drive |
|||
[1] See the graphics boards and driver requirements for Open Inventor at https://developer.openinventor.com.
Attachment(s):
| File | Last Modified |
|---|---|
| 269-366900A_OMNIC Paradigm Tiers.pdf | November 05, 2024 |