OMNIC Picta Release Notes
About This Document
This document contains a revision history of OMNIC Picta, including new features that may not be included in the User’s Guide, resolved issues, and known issues.
Trademarks
Microsoft, Internet Explorer, and Windows are either trademarks or registered trademarks of Microsoft Corporation in the United States and/or other countries.
ToolBook is either a trademark or registered trademark of SumTotal in the United States and/or other countries.
Semprex is either a trademark or registered trademark of Semprex Corporation in the United States and/or other countries.
PerkinElmer is either a trademark or registered trademark of PerkinElmer, Inc. in the United States and/or other countries.
Sadtler and SearchMaster are trademarks of Bio-Rad Laboratories, Inc. in the United States and/or other countries.
Bruker and OPUS are either trademarks or registered trademarks of Bruker Optics, Inc. in the United States and/or other countries.
All other trademarks are the property of Thermo Fisher Scientific Inc. and its subsidiaries.
|
Note
|
See the OMNIC and OMNIC Atlµs Release Notes for information about changes to those applications that may affect OMNIC Picta. |
OMNIC Picta 1.9 Hotfix 1
-
Build number: 1.9.1.22
-
Release date: June 2025
-
Supported operating systems
-
Windows 10 64-bit Version 1607 and newer
-
Windows 11 64-bit
-
New Features
Support new Sentech Camera
iN10 Diagnostics has been updated to properly support the new USB3 Sentech camera.
Resolved Issues
None
Known Issues
None
OMNIC Picta 1.9
Release Date: Dec-2023
Supported Operating Systems
-
Windows 10 64-bit
-
Windows 11 64-bit
New Features
None
Resolved Issues
OMNIC Picta Does Not Prompt for Spectrum Title Before Single Sample Data Collection
When collecting a single sample scan, the software does not prompt the user to enter a spectrum title before collecting the spectrum. This causes issues when the "Auto save a collected spectrum using the spectrum title" Data Security policy is enabled. The spectrum will be saved with the default title, which is the timestamp.
Known Issues
None
OMNIC Picta 1.8 Hotfix 2
Release Date: Apr-2022
Supported Operating Systems
-
Windows 10 64-bit
-
Windows 11 64-bit
New Features
None
Resolved Issues
Crash when applying flat field correction to mosaics
When a mosaic is collected or displayed with flat field correction enabled, an "Unhandled Exception" dialog appears.
Known Issues
None
OMNIC Picta 1.8 Hotfix 1
Release Date: Feb-2022
Supported Operating Systems
-
Windows 10 64-bit
New Features
Update drivers for new core card
This release updates drivers needed for the iN10 core card update.
Resolved Issues
OMNIC Specta crashes when performing spectral interpretation
With OMNIC Picta installed, OMNIC Specta program crashes when doing the spectral interpretation and during the multi component search.
Deleting detector spectral range causes unhandled exception on active detector
Completely deleting detector spectral range causes unhandled exception on active detector when trying to change the min. spectral range.
Data Security - Open experiment file allows changing directory
When the "Prevent changing directories when opening files" policy is set, the user can change directories when opening an experiment file. However, you cannot change directories when saving an experiment file.
Data Security - Choosing to use a saved background allows changing directory
When the "Prevent changing directories when opening files" policy is set, the user can change directories when choosing to use a saved background.
Known Issues
Crash when applying flat field correction to mosaics
When a mosaic is collected or displayed with flat field correction enabled, an "Unhandled Exception" dialog appears.
OMNIC Picta 1.8
Release Date: May-2021
Supported Operating Systems
-
Windows 10 64-bit
New Features
Add option for automatic baseline correction during map collection
The ability to do automatic baseline correction on each spectrum during a collect has been added.
Add option for automatic atmospheric correction during map collection
The ability to do automatic atmospheric correction on each spectrum during a collect has been added.
Particle analysis file for analyze
When the particle analysis wizard is complete, the data can be saved as a map file. A new file will also be saved the will allow the reanalysis of the particle map in OMNIC Atlµs.
Resolved Issues
Crashes after multiple uses of particle wizard
OMNIC Picta has crashed on some occasions when multiple particle wizard runs are done sequentially with a large number of particles.
Known Issues
Deleting detector spectral range causes unhandled exception on active detector
Completely deleting detector spectral range causes unhandled exception on active detector when trying to change the min. spectral range.
Data Security - Open experiment file allows changing directory
When the "Prevent changing directories when opening files" policy is set, the user can change directories when opening an experiment file. However, you cannot change directories when saving an experiment file.
Data Security - Choosing to use a saved background allows changing directory
When the "Prevent changing directories when opening files" policy is set, the user can change directories when choosing to use a saved background.
Crash when applying flat field correction to mosaics
When a mosaic is collected or displayed with flat field correction enabled, an "Unhandled Exception" dialog appears.
OMNIC Picta 1.7 Hotfix 6
Release Date: July-2020
Supported Operating Systems
-
Windows 10 64-bit
New Features
None
Resolved Issues
Data Security - Collect spectrum does not always save
Before collecting a map, collecting a spectrum will save and prompt for a signature. After collecting a map, then collecting a spectrum, the spectrum will not automatically save and prompt for signature.
Data Security - Particle wizard crash
After using the particle wizard several times, it will crash.
Data Security - Collect engine does not start
At times the collect engine does not start when OMNIC Picta is started. The wait time for the collect engine has been increased to allow for long delays on distributed security systems.
Known Issues
Deleting detector spectral range causes unhandled exception on active detector
Completely deleting detector spectral range causes unhandled exception on active detector when trying to change the min. spectral range.
OMNIC Picta 1.7 Hotfix 5
Release Date: May-2020
Supported Operating Systems
-
Windows 10 64-bit
New Features
None
Resolved Issues
Data Security - Particle analysis file sign
When collecting a map with more than 100 particles the software would crash after attempting to sign the map file.
Known Issues
None
OMNIC Picta 1.7 Hotfix 4
Release Date: April-2020
Supported Operating Systems
-
Windows 10 64-bit
New Features
None
Resolved Issues
Data Security - Particle analysis file save
When saving a file after a particle analysis, an existing file could be overwritten even though the policy defined this as not allowed.
Data Security - Particle analysis file signing
When saving a file after a particle analysis, the file is signed. When verifying the signature, the file is reported as being modified since it was signed. The file is now correctly signed.
Known Issues
None
OMNIC Picta 1.7 Hotfix 3
Release Date: February-2020
Supported Operating Systems
-
Windows 10 64-bit
New Features
Data Security - Particle wizard collect cancel
The particle wizard has been enhanced to not allow cancelation of the wizard during all the collection and process steps until the data has been saved. When cancel is not allowed, the process stage will force a file save to a map file of all data so that it will be properly saved and signed.
Resolved Issues
Data Security - Save map to secure folder
OMNIC Picta will now save maps only to the secure folder when security policy defines that state. This has been improved to support all security cases.
Known Issues
None
OMNIC Picta 1.7 Hotfix 2
Release Date: November-2019
Supported Operating Systems
-
Windows 10 64-bit
New Features
Data Security - Report printed before saving data
Reports could be printed without saving data. This has been changed to prevent printing until the data is saved and signed.
Data Security - Save map to secure folder
OMNIC Picta will now save maps only to the secure folder when security policy defines that state.
Data Security - Add audit events
Audit events have been added to several actions. These include print from particle wizard, save video image, print from the file menu, save profile as CSV and print button in analyze images option.
Resolved Issues
Imaging detector not available
When using the software in French, the imaging detector was not available.
Some issues in German
The stage movement and mosaic did not operate correctly in German.
Known Issues
None
OMNIC Picta 1.7 Hotfix 1
Release Date: February-2019
Supported Operating Systems
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
New Features
None
Resolved Issues
Particle analysis only reports 100 particles
When collecting a map with many particles, only 100 are reported.
Particle analysis background button disabled
When setting up a particle analysis map collect, the background step was blocked by a disabled button.
Changing operating system region loses video calibration
When video has been calibrated and then the region information is changed in the operating system so that decimal point type is not correct, the video calibration is lost. The fix for this is to edit the OMNIC_MC.INI file. Change the decimal point in the calibration section to use either a comma or period to match the local settings then the calibration can be used.
Particle analysis image analysis needs restart
When doing an image analysis report, the software would crash when multiple items clicked in table.
Maps do not collect when using security suite
When starting a map collect with security setup, the maps would not collect.
Particle analysis bar graph
Sometimes the bar graph for size distribution would not match the actual size distribution.
Known Issues
None
OMNIC Picta 1.7
Release Date: October-2018
Supported Operating Systems
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
New Features
Software now reports length and width of particle size
When doing the particle wizard, it now reports the particle length and width.
Resolved Issues
Spectra in particle analysis report not scaled to full size
The spectra in the particle analysis report are shown as common scaled so if one spectrum is large compared to the others, the other spectra will be just a line. The report has been changed to show all spectra at full scale.
Problem when changing between the virtual and mechanical joystick on iN10
The OMNIC Picta stage tool is not synchronized with the instrument stage when changing between the virtual and the mechanical joystick.
Software becomes unstable when collecting or processing large map or mosaics
Extensive changes to improve memory usage, user warnings and error trapping when collecting larger maps and mosaics.
Software becomes unresponsive when collecting large ATR maps
Fixed an issue that caused memory loss when collecting large ATR maps, eventually causing the collect to stop.
Fixed issue with OMNIC Picta not starting
Fixed an issue that prevented OMNIC Picta from starting intermittently.
Known Issues
None
OMNIC Picta 1.6 Hotfix 2
Release Date: May-2018
Supported Operating Systems
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
New Features
None
Resolved Issues
Crash when using the Analyze Images tool
A crash was found when using the image analysis tool when selecting the second step.
Unable to save particle analysis results in a map
When attempting to save particle analysis results to a map, the file was not created.
Known Issues
None
OMNIC Picta 1.6 Hotfix 1
Release Date: Oct-2017
Supported Operating Systems
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
New Features
None
Resolved Issues
Spectrum header shows incorrect experiment filename
When viewing a spectrum header for a spectrum collected in OMNIC Picta, it shows the incorrect experiment filename. This has been changed to show the OMNIC Picta experiment filename.
Known Issues
None
OMNIC Picta 1.6
Release Date: May-2017
Supported Operating Systems
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
New Features
Added Data Security (DS) features
-
Add policy to prevent cancel of sample collection
-
Add policy to auto save spectra
-
Add policy to control use of Cancel button on signing dialog
-
Add audit event for start of a collection
-
Add audit event when cancel login to OMNIC Picta
-
At the start of the application, the status of the security suite will be checked and report any issues
-
Instrument information is now available for reports
-
The signature dialog will not stop after 3 fails. It will continue to remain open
Resolved Issues
Login fail will open application in DS mode
The application will not start after failed logins. It will record an event in the event log
Particle Wizard reports 0% area
Under certain conditions, a particle was reported as 0%.
Cannot save experiment files in non-English languages
Certain parameters were not persisted when saved in languages other than English.
Advanced diagnostic reports wrong instrument name
When doing a report in the advanced diagnostic test, the wrong instrument name was reported.
Save map not saving all data
It was found that when processing a map of discrete map points, the changed data could not be saved. This will now function correctly and save the new data and the information about the spectral history.
ATR release distance problem
When set to languages other than English, the ATR release distance was not always the selected value.
ATR use of background reference
When loading an experiment file saved with the ATR collect mode, the background reference button was not disabled for selection. All background buttons are not in the correct state when the experiment file is loaded.
Aperture settings not saved
The aperture settings were not saved in the map file. Therefore, the map information did not report the settings at collection. This has been changed to save the aperture settings.
Known Issues
None
OMNIC Picta 1.5 Hotfix 2
Release Date: Aug-2016
Supported Operating Systems
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
New Features
None
Resolved Issues
Mosaic for line maps not always correct
When collecting a line map, drawing the line from left to right or right to left will affect how the mosaic is collected.
Multiple Save Map commands shown
After collecting a map, there can be multiple Save Map commands shown.
Export map after normalizing scale
When exporting a map after a normalize scale operation, the file will not always be saved.
OMNIC Picta stops after spectrum copy
After selecting a point on a point map and copying the spectrum, OMNIC Picta would hang up.
OMNIC Picta particle wizard hangs up
When using an OMNIC Picta particle wizard, the software would sometimes hang up.
Known Issues
None
OMNIC Picta 1.5 Hotfix 1
Release Date: Oct-2015
Supported Operating Systems
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
New Features
None
Resolved Issues
DS mode allows use of software when defined as disabled
When Thermo Security is set up to not allow certain users to open OMNIC Picta, OMNIC Picta would put a message that stated it would not open but then went and opened anyway.
Known Issues
None
OMNIC Picta 1.5
Release Date: Oct-2014
Supported Operating Systems
-
Windows XP SP3 32-bit
-
Windows 7 32-Bit
-
Windows 7 64-bit
New Features
ATR release distance
Increased distance stage moves down when releasing ATR.
Translations
Polish translation is complete, improved Japanese and German translations.
Resolved Issues
Discrete map "Go to point" issue
Fixed issue when setting up discrete point maps and attempting to use the "Go to point" on the first point.
Background collect after time issue
Fixed to collect background after time so that new background is collected at the specified time.
Use saved background issue
Saved backgrounds were not being used, instead used in memory background.
Saving profiles in map issue
New profiles were not saved to some maps. The profile will now be saved to the map file.
OMNIC Picta did not use OMNIC DS settings issue
OMNIC Picta will now use the settings from OMNIC DS that are appropriate.
OMNIC Picta logging failed logins
OMNIC Picta will now log failed logins so that they can be monitored.
OMNIC Picta auto focus
OMNIC Picta auto focus has been improved.
Too short a time between sample and background collects
The time between sample and background collects has been increased. It is also adjustable using a setting at the bottom of the experiment setup page.
Could not apply changes to maps
Operations to change format, apply Kramer’s Kronig or other operations on a map have been fixed to work with all maps.
Known Issues
None
OMNIC Picta 1.4
Release Date: Jul-2012
Supported Operating Systems
-
Windows XP SP3 32-bit
-
Windows 7 32-Bit
-
Windows 7 64-bit
New Features
ATR release distance
You can now specify an additional release distance with the ATR pressure profile.
Background handling
Collected backgrounds are not invalidated by changing collection parameters and reverting to previous values.
Background handling
Collecting background after sample has been disabled for ATR data collection.
Application Support
OMNIC Picta has been updated to support features of OMNIC 9.
OMNIC Picta Dialogs
Changes have been made in OMNIC Picta so that dialog boxes will not drop behind the application or become forced topmost windows.
Resolved Issues
None
Known Issues
Collect Background after sample
Collect Background after sample does not work for maps and this feature will be disabled. This feature will be re-enabled when fixed.
OMNIC Picta 1.3
Release Date: Oct-2011
Supported Operating Systems
-
Windows XP SP3 32-bit
-
Windows 7 32-Bit
-
Windows 7 64-bit
New Features
None
Resolved Issues
64bit support
OMNIC Picta now supports 64-bit Windows.
OMNIC Picta Dialogs
Changes have been made in OMNIC Picta so that dialog boxes will not drop behind the application.
Known Issues
None
OMNIC Picta 1.2 Hotfix 2
Release Date: Jun-2011
Supported Operating Systems
-
Windows XP SP3 32-bit
-
Windows 7 32-Bit
New Features
None
Resolved Issues
Live Video image shrinks during use
If you have a manual XY stage, the live video image may shrink during use. The only way to resize the live video image was to restart OMNIC Picta. If you have a motorized XY stage, you are not affected by this issue and no additional fixes are included.
Known Issues
None
OMNIC Picta 1.2 Hotfix 1
Release Date: Feb-2011
Supported Operating Systems
-
Windows XP SP3 32-bit
-
Windows 7 32-Bit
New Features
None
Resolved Issues
Analyze Images produces exception Error
When using the Image Analysis tool in OMNIC Picta an exception error will be displayed and the feature will work.
Point map displays Single Beams instead of ratioed spectra
When collecting a Point map with multiple aperture sizes, a single beam may be displayed instead of ratioed spectra.
DDiag indicates unable to communicate
When using DDiag to communicate with the iN10, it may indicate a failure to communicate and not allow the requested operation.
Incorrect Z axis location used with saved Point Maps
When a Point map experiment is saved and reloaded at a later time, the original Z axis values of each sample point is not correctly restored.
| To update an existing experiment file with Z axis information, load the experiment file, "touch" each sample point to update its X, Y and Z coordinate and then save the experiment. |
Known Issues
None
OMNIC Picta 1.2
Release Date: Jun-2010
Supported Operating Systems
-
Windows XP SP3 32-bit
New Features
Digital Security
OMNIC Picta now supports digital security and the Thermo Security Administration application.
Default Experiment
A user may choose to select a specific default OMNIC Picta experiment file to be loaded at startup. To select a default experiment file, navigate to the Advanced section of Experiment Setup in the View and Collect section.
Instrument Firmware version
The instrument firmware version can be found by opening the About OMNIC Picta dialog in the Help section.
Resolved Issues
None
Known Issues
None
OMNIC Picta 1.1
Release Date: Sep-2009
Supported Operating Systems
-
Windows XP SP3 32-bit
New Features
Map Math
When two like maps are opened in OMNIC Atlµs the Map Math function is available. The Map Math function provides the following map math functionality
(Map 1 = A and Map 2 = B and k = constant):
A/B
B/A
A*B
B-A
A+kB
A-+kB
(A-kB) / (A+kB)
Send to OMNIC Specta
A right-click option has been added to send map spectra to the OMNIC Specta application as temporary files.
Mosaic has not changed
If the mosaic has not changed since the last time collected prior to map collection, it will not be recollected. Instead the current mosaic collection will be used for the map collection.
DDiag now has live video image display
DDiag now has a live video image display.
Progress indicator now available during optimize condenser
A progress window is now available during the optimize condenser operation.
Stop button for optimize signal process
A stop button is now available during the optimize signal operation.
Resolved Issues
Overlay of chemical image and video image in transmission is working correctly
The Map Blend option did not work correctly when in transmission.
Cross section and Random Mixture wizard color selection is not correct
When selecting color options in the Cross section and Random Mixture wizards did not work correctly in version 1.0.
Data collection with a manual stage
Data collection now works properly and waits correctly between the background and sample collection.
Condenser position is retained
When setting a new background, the condenser position is now remembered.
Changing the aperture size invalidates the background
Changing the aperture size now invalidates the background.
Additional experiment settings are retained
When reopening the OMNIC Picta application, additional experiment settings are now retained such as background collection settings, number of scans, resolution, number of background scans, aperture size, final format, step size, apodization and scan speed.
ATR mode switching
Only the data format is changed when changing the data format in ATR mode, the ATR pressure is no longer released and reapplied.
System Status for Electronics spawns multiple windows
Multiple windows are no longer spawned when viewing the Electronics system status.
Ultra-fast mapping is now available after using Array Zoom
Ultra-fast mapping option is now available after using the array zoom feature.
Drawing area maps at the viewing limits is now acceptable
Drawing area maps at the viewing limits was not allowed in version 1.0 of OMNIC Picta.
Array zoom now requires a new background
Array zoom now requires a new background collection.
DDiag window issues have been addressed
DDiag window issues have been addressed such as windows appearing behind other windows.
ATR release distance has been increased
The ATR release distance has been increased from the previous version.
Rotating the aperture by entering a number into the dialog now works
In the previous version of the software rotating the aperture by modifying the values in the dialog boxes now work correctly along with rotating the rectangle in the window.
Macro library setup option is now working
The macro library setup option is now available in OMNIC Picta.
Backup parameters can now be saved in DDiag
DDiag now supports and option to save the backup parameters.
Translation Updates
The translation files have been updated.
Known Issues
None
OMNIC Picta 1.0
Release Date: May-2008
Supported Operating Systems
-
Windows XP SP3 32-bit
New Features
Initial Release, all features are new
Resolved Issues
Initial release, there are no resolved Issues
Known Issues
QC Libraries
QC libraries are not available in OMNIC Picta. However, QC library options do appear in Library Manager and Library Setup
Close or Minimize
Close or minimize all applications when running OMNIC Picta. At times if other applications are running behind OMNIC Picta, they can show through the application, and spectral or map data windows may not appear correctly in Analyze Maps and Data.
Area Drawing
When drawing an area map with the array mode selected, use the Mosaic pane to specify the area rectangle instead of the live video pane, because the live video pane may cause the display to jump
TQ Analyst
TQ Analyst must be reinstalled after OMNIC Picta if it existed on the system before OMNIC Picta installation. The OMNIC Picta installer will install TQ EZ instead.
Line Segments
Line segments larger than half the current step size must be specified for the line segment to be drawn on the display.
Photoacoustic
Photoacoustic processed data does not work well with the wizards.
Thermo IQ
When you are running the Thermo Software IQ program, a warning will appear indicating that the omnic_wc.dll installed with OMNIC Picta is a newer version than expected for OMNIC. This is expected with the OMNIC Picta version 1.0 release. There will also be a warning message because the OMNICAdministrator.exp file size is different when installed with the iN10 driver. This is an obsolete file that is no longer used, and the change in file size will not affect the software functionality.
How to Contact Us
Current contact information is located at https://www.thermofisher.com
Select the "Contact Us" icon at the top of the screen
Attachment(s):
| File | Last Modified |
|---|---|
| OMNIC Picta Release Notes.pdf | January 13, 2026 |