269-3601 Using Spectral Math
This tutorial explains how to use the Spectral Math function in OMNIC software to perform arithmetic operations on spectra. Users can subtract, add, divide, or scale spectra to correct baselines, compare materials, or create synthetic spectra for analysis and identification.
Description:
This tutorial explains how to use the Spectral Math command in the Process menu of your OMNIC software to perform arithmetic and other math operations on spectra.
Click the buttons at the left to view the lessons. The Overview introduces you to the features of the Spectral Math window. The other lessons are examples that take you through ..
- Ratioing two spectra: A/ B
- Creating a synthetic spectrum using a factor: A + k•B
Environment:
OMNIC Software
Attachment(s):
| File | Last Modified |
|---|---|
| 269-3601 Using Spectral Math.pdf | December 02, 2022 |