Renaming spectral libraries in OMNIC
Omnic does not support directly renaming spectral libraries, but users can recreate and merge libraries using compatible settings. Field mismatches may cause data loss during the merge process.
Description:
OMNIC does not have a direct way to change the name of a Search or QC library. The workaround is to create a new library with the same set of custom information fields and spectral range as the original library. The attached document describes the process for creating the new library.
Environment:
OMNIC
Attachment(s):
| File | Last Modified |
|---|---|
| Renaming spectral libraries in OMNIC.pdf | December 16, 2021 |