OMNICxi Optical Component Files Release Notes
About This Document
This document contains a revision history of OMNICxi Optical Component Files, including new features that may not be included in the User’s Guide, resolved issues, and known issues.
Trademarks
Microsoft, Internet Explorer, and Windows are either trademarks or registered trademarks of Microsoft Corporation in the United States and/or other countries.
ToolBook is either a trademark or registered trademark of SumTotal in the United States and/or other countries.
Semprex is either a trademark or registered trademark of Semprex Corporation in the United States and/or other countries.
PerkinElmer is either a trademark or registered trademark of PerkinElmer, Inc. in the United States and/or other countries.
Sadtler and SearchMaster are trademarks of Bio-Rad Laboratories, Inc. in the United States and/or other countries.
Bruker and OPUS are either trademarks or registered trademarks of Bruker Optics, Inc. in the United States and/or other countries.
All other trademarks are the property of Thermo Fisher Scientific Inc. and its subsidiaries.
OMNICxi Optical Component Files v1.2
Release Date: May-2020
Supported Operating Systems
Windows 7 32-Bit
Windows 7 64-bit
Windows 8.1 64-bit
Windows 10 64-bit
New Features
Add support for other gratings for 532 and 633 high power lasers
Previously only the full range gratings worked for the new high power 532 and 633 lasers. Now there is support for the high res and extended range grating for 532, and high res for the 633 laser.
Resolved Issues
None
Known Issues
None
OMNICxi Optical Component Files v1.1.5
Release Date: Aug-2019
Supported Operating Systems
Windows 7 32-Bit
Windows 7 64-bit
Windows 8.1 64-bit
Windows 10 64-bit
New Features
Add 532 and 633 high power lasers
The parameters for the 532 and 633 high power lasers were added..
Resolved Issues
None
Known Issues
None
OMNICxi Optical Component Files v1.1
Release Date: Jan-2018
Supported Operating Systems
Windows 7 32-Bit
Windows 7 64-bit
Windows 8.1 64-bit
Windows 10 64-bit
New Features
None
Resolved Issues
Limits for the 3054 polystyrene peak expanded
Performance check limits after align and calibrate were narrower than the tightest pharmacopeia values. The 3054 polystyrene peak is a cluster of peaks and this causes more variation in peak position than other single peaks in the spectrum. The new limits match the pharmacopeia limits for that peak.
Known Issues
None
OMNICxi Optical Component Files v1.0
Release Date: Dec-2016
Supported Operating Systems
Windows 7 32-Bit
Windows 7 64-bit
Windows 8.1 64-bit
Windows 10 64-bit
New Features
Initial Release, all features are new
Resolved Issues
Initial Release, all features are new
Known Issues
None
How to Contact Us
Current contact information is located at https://www.thermofisher.com
Select the "Contact Us" icon at the top of the screen
Attachment(s):
| File | Last Modified |
|---|---|
| OMNICxi optical component files release notes.pdf | March 20, 2024 |