Backing up OMNIC Paradigm database to a Network Location
Description:
The OMNIC Paradigm software has a database backup and restore function accessible from the Configure menu in the Desktop Mode. The Backup function allows the user to specify a folder for where the backup will be created. This can be a local folder on the Summit computer, a USB drive connected to the computer, or to a network location if the computer is connected to the customer’s network.
Environment:
- OMNIC Paradigm
- Nicolet Summit
Attachment(s):
File | Last Modified |
---|---|
Backing up OMNIC Paradigm database to a network location.pdf | March 08, 2022 |