Security Suite Release Notes
Security Suite Release Notes
About This Document
This document contains a revision history of Security Suite, including new features that may not be included in the User’s Guide, resolved issues, and known issues.
The following conventions are used in this document:
|
Note
|
Brief information or reminder. |
|
Tip
|
Helpful suggestion or advice. |
|
Warning
|
A situation that could result in damage to system or data loss. |
Trademarks
Microsoft, Internet Explorer, and Windows are either trademarks or registered trademarks of Microsoft Corporation in the United States and/or other countries.
ToolBook is either a trademark or registered trademark of SumTotal in the United States and/or other countries.
Semprex is either a trademark or registered trademark of Semprex Corporation in the United States and/or other countries.
PerkinElmer is either a trademark or registered trademark of PerkinElmer, Inc. in the United States and/or other countries.
Sadtler and SearchMaster are trademarks of Bio-Rad Laboratories, Inc. in the United States and/or other countries.
Bruker and OPUS are either trademarks or registered trademarks of Bruker Optics, Inc. in the United States and/or other countries.
All other trademarks are the property of Thermo Fisher Scientific Inc. and its subsidiaries.
General Notes
-
Audit Manager, the Security Suite services, and the Security Suite client applications all generate operational log files. These files are not required for the audit trail and are intended for troubleshooting purposes; as such, they can be deleted if necessary to recover disk space. The log files are located in the following folders:
-
%AppData%\Thermo Scientific\AuditManager– Audit Manager 1.9 and older; each user will have their own log files -
%ProgramData%\Thermo Scientific\AuditEventWriterService -
%ProgramData%\Thermo Scientific\Audit Manager– Audit Manager 2.7 and newer -
%ProgramData%\Thermo Scientific\Security Suite
-
-
Please see Computer Requirements for FTIR and Raman Instruments for the supported operating systems and databases.
Security Suite 2.7 Hotfix 2
-
Build number: 2.7.2.33
-
Release date: March 2026
-
Supported operating systems
-
Server
-
Windows Server Standard with Desktop Experience 2016 and newer
-
Windows Server Datacenter with Desktop Experience 2016 and newer
-
-
Workstation
-
Windows 10 64-bit Version 1607 and newer
-
Windows 11 64-bit
-
-
New Features
None
Resolved Issues
Unable to sign a spectrum in OMNIC and OMNIC for Dispersive Raman
If the user simply opens a spectrum in OMNIC and attempts to sign it, an error message indicating that it is unable to write to the audit log and the operation cannot proceed.
Incorrect error when launching OMNIC or OMNIC for Dispersive Raman
If the "Prevent changing directories when saving files" policy is not enabled and the NTFS permissions for the directory for spectral data did not permit the current user to write files, an error message is displayed when launching OMNIC or OMNIC for Dispersive Raman.
Service Startup Type Set to "Automatic (Delayed Start)"
The Thermo Scientific Secure Data Storage Service, Thermo Scientific File Change Monitor Service, and Thermo Scientific Security Service startup types were set to "Automatic (Delayed Start)" during installation or upgrade. This caused issues with launching applications that rely on these services immediately after system startup.
Accounts blank passwords are not supported
User accounts with blank passwords were not supported for use with Security Suite 2.7 and 2.7 Hotfix 1.
User authentication issues for local accounts
Due to the changes to support Kerberos authentication, local accounts could be authenticated improperly in some situations.
Unable to open Security Administration on non-English systems
Due to a translation issue, Security Administration would fail to open on non-English systems.
Passwords with special characters could cause installation to fail
If the password for the Data Storage Service contained double quotation marks ("), the installation would fail to launch.
The File Change Monitor Service could stop unexpectedly
If a file located in a shared directory was modified, deleted, etc., the Thermo Scientific File Change Monitor Service could stop unexpectedly when the resulting UNC path name exceeded 259 characters.
Blank Warning Message Appears When Selecting a UNC Path for Secure Root Folder
If you select a UNC path for the secure root folder in Security Administration, a blank warning message box appears. The erroneous message box can be closed safely.
Incorrect Passwords Cause Premature Account Locking
Entering an incorrect password for Security Suite applications can cause account lockout to occur after fewer attempts than the computer policies indicate.
Known Issues
Excluding directories from file monitoring does not work as expected
There are multiple limitations when excluding directories from file monitoring in Security Administration:
-
Wildcards in the middle of a directory name are not supported and will not work as expected. For example, excluding
Temp*Dirwill actually exclude all files in any folder containingTempDirin their name, such asMyTempDirFolderorTempDirectory. -
There is an implied leading and trailing asterisk (\*) wildcard for each directory name. For example, excluding
Tempwill actually exclude all files in any folder containingTempin their name, such asMyTempFolderorTemporaryFiles. To only exclude files in a folder named exactlyTemp, you must specify\Temp\(including the backslashes). -
The question mark (?) wildcard is not supported at all. Since the question mark is not a valid character in a directory name on Windows systems, attempting to use it will not exclude any directories.
-
The directory name should not include the drive letter or root backslash. For example, to exclude all files in
C:\Temp, you should only enterTemp\.
Unable to install on some network configurations
Attempting to install Security Suite using a domain account for the Data Storage Service or Audit Log Service credentials will fail if the computer is not joined to the same domain or another trusted domain in the same forest. This is due to how Windows handles authentication for services.
- Workaround
-
Join the computer to an appropriate domain before installing Security Suite.
Security Suite 2.7 Hotfix 1
-
Build number: 2.7.1.11
-
Release date: December 2025
-
Supported operating systems
-
Server
-
Windows Server Standard with Desktop Experience 2016 and newer
-
Windows Server Datacenter with Desktop Experience 2016 and newer
-
-
Workstation
-
Windows 10 64-bit Version 1607 and newer
-
Windows 11 64-bit
-
-
New Features
None
Resolved Issues
Incorrect secure data directory value
Security Suite 2.7 failed to replace the environment variable in the secure data directory policies when returning the value to applications. This results in the system environment variable being used instead of the value specified in Security Administration.
Log files for OMNIC and OMNIC for Dispersive Raman filled with nuisance warnings
Due to how OMNIC and OMNIC for Dispersive Raman (OFDR) handle file I/O, a large number of warning messages were logged in the Security Suite log files when these applications were used. These warnings were generated when the application closed files it had opened for reading since Security Suite always called a function that only applied to files opened for writing which would generate a warning. This function call has been removed for files opened for reading to prevent these warnings from being logged.
Known Issues
Excluding directories from file monitoring does not work as expected
There are multiple limitations when excluding directories from file monitoring in Security Administration:
-
Wildcards in the middle of a directory name are not supported and will not work as expected. For example, excluding
Temp*Dirwill actually exclude all files in any folder containingTempDirin their name, such asMyTempDirFolderorTempDirectory. -
There is an implied leading and trailing asterisk (\*) wildcard for each directory name. For example, excluding
Tempwill actually exclude all files in any folder containingTempin their name, such asMyTempFolderorTemporaryFiles. To only exclude files in a folder named exactlyTemp, you must specify\Temp\(including the backslashes). -
The question mark (?) wildcard is not supported at all. Since the question mark is not a valid character in a directory name on Windows systems, attempting to use it will not exclude any directories.
-
The directory name should not include the drive letter or root backslash. For example, to exclude all files in
C:\Temp, you should only enterTemp\.
Accounts blank passwords are not supported
Due to how the user authentication protocols work, accounts with blank passwords are not supported for use with Security Suite.
Unable to install on some network configurations
Attempting to install Security Suite using a domain account for the Data Storage Service or Audit Log Service credentials will fail if the computer is not joined to the same domain or another trusted domain in the same forest. This is due to how Windows handles authentication for services.
- Workaround
-
Join the computer to an appropriate domain before installing Security Suite.
Unable to sign a spectrum in OMNIC and OMNIC for Dispersive Raman
If the user simply opens a spectrum in OMNIC and attempts to sign it, an error message indicating that it is unable to write to the audit log and the operation cannot proceed.
Incorrect error when launching OMNIC or OMNIC for Dispersive Raman
If the "Prevent changing directories when saving files" policy is not enabled and the NTFS permissions for the directory for spectral data did not permit the current user to write files, an error message is displayed when launching OMNIC or OMNIC for Dispersive Raman.
Service Startup Type Set to "Automatic (Delayed Start)"
The Thermo Scientific Secure Data Storage Service, Thermo Scientific File Change Monitor Service, and Thermo Scientific Security Service startup types were set to "Automatic (Delayed Start)" during installation or upgrade. This caused issues with launching applications that rely on these services immediately after system startup.
Unable to open Security Administration on non-English systems
Due to a translation issue, Security Administration would fail to open on non-English systems.
Security Suite 2.7
-
Build number: 2.7.0.172
-
Release date: November 2025
-
Supported operating systems
-
Server
-
Windows Server Standard with Desktop Experience 2016 and newer
-
Windows Server Datacenter with Desktop Experience 2016 and newer
-
-
Workstation
-
Windows 10 64-bit Version 1607 and newer
-
Windows 11 64-bit
-
-
Important Changes
-
As of this release, the Audit Manager release notes have been combined with the Security Suite release notes. Additionally, the version number for the Audit Log Service and Audit Manager applications now match the Security Suite version number.
-
The Security Suite documentation installer has been retired. The latest documentation is now available online at Security Suite Manuals.
-
The Security Suite Language Pack installer has been merged into the main Security Suite installer. During an upgrade, the Security Suite Language Pack will be uninstalled and the main installer will install the language files automatically.
New Features
Added support for Kerberos authentication for applications using the COM security client
Updated the user authentication code to support Kerberos authentication when the LDAP server supports it. This only applies to applications using the COM security client, such as OMNIC or RESULT. Applications using the .NET security client, such as Audit Manager and OMNIC Paradigm, already support Kerberos authentication.
Resolved Issues
File creation and modification events are not properly logged in Security Suite 2.6 Hotfix 2
Due to errors in the file monitoring code, some file extensions were improperly excluded from creation and modification events. This has been fixed so that all file types are properly monitored.
Improved memory usage
Improved the memory usage of the COM client, Security Administration, and the Security service to minimize the amount of memory used which improves system stability and performance.
Upgrading Security Suite on systems with OMNIC Specta gives an error message during installation
When upgrading Security Suite on a system that has OMNIC Specta installed and the Thermo Indexer running, you may receive an error message after entering the username and password to use for the Secure Data Storage service.
Known Issues
Excluding directories from file monitoring does not work as expected
There are multiple limitations when excluding directories from file monitoring in Security Administration:
-
Wildcards in the middle of a directory name are not supported and will not work as expected. For example, excluding
Temp*Dirwill actually exclude all files in any folder containingTempDirin their name, such asMyTempDirFolderorTempDirectory. -
There is an implied leading and trailing asterisk (\*) wildcard for each directory name. For example, excluding
Tempwill actually exclude all files in any folder containingTempin their name, such asMyTempFolderorTemporaryFiles. To only exclude files in a folder named exactlyTemp, you must specify\Temp\(including the backslashes). -
The question mark (?) wildcard is not supported at all. Since the question mark is not a valid character in a directory name on Windows systems, attempting to use it will not exclude any directories.
-
The directory name should not include the drive letter or root backslash. For example, to exclude all files in
C:\Temp, you should only enterTemp\.
Accounts blank passwords are not supported
Due to how the user authentication protocols work, accounts with blank passwords are not supported for use with Security Suite.
Unable to install on some network configurations
Attempting to install Security Suite using a domain account for the Data Storage Service or Audit Log Service credentials will fail if the computer is not joined to the same domain or another trusted domain in the same forest. This is due to how Windows handles authentication for services.
- Workaround
-
Join the computer to an appropriate domain before installing Security Suite.
Incorrect secure data directory value
Security Suite 2.7 fails to replace the environment variable in the secure data directory policies when returning the value to applications. This results in the system environment variable being used instead of the value specified in Security Administration.
- Workaround
-
Update the value of the environment variable(s) for the user to the secure data folder value.
WarningThis may have further implications if other applications rely on the environment variable, so careful testing of the change is advised.
Service Startup Type Set to "Automatic (Delayed Start)"
The Thermo Scientific Secure Data Storage Service, Thermo Scientific File Change Monitor Service, and Thermo Scientific Security Service startup types were set to "Automatic (Delayed Start)" during installation or upgrade. This caused issues with launching applications that rely on these services immediately after system startup.
Unable to open Security Administration on non-English systems
Due to a translation issue, Security Administration would fail to open on non-English systems.
Security Suite 2.6 Hotfix 2
-
Build number: 2.6.2.17
-
Release date: August 2024
-
Supported operating systems
-
Server
-
Windows Server 2016 w/ Desktop Experience
-
Windows Server 2019 w/ Desktop Experience
-
Windows Server 2022 w/ Desktop Experience
-
-
Workstation
-
Windows 10 64-bit Version 1607 and newer
-
Windows 11 64-bit
-
-
New Features
None
Resolved Issues
Unable to add users or groups to Access Control items
When adding users or groups to Access Control items, the user interface would stop responding and never recover.
Erroneous message when trying to save configuration files in OMNIC and OMNIC for Dispersive Raman
When trying to save configuration files in OMNIC and OMNIC for Dispersive Raman, a message would appear stating that the audit log could not be written to, and the operation could not be performed without logging.
The Thermo Scientific Secure Data Storage Service would stop when the Thermo Scientific Security Service was restarted
When the Thermo Scientific Security Service was restarted (e.g. as part of reconfiguring the audit log database), the Thermo Scientific Secure Data Storage Service would stop and not restart automatically.
Automatic dump file creation did not work properly on 64-bit systems
Due to how Microsoft Windows handles 32-bit applications on 64-bit systems, the automatic creation of dump files did not work properly on 64-bit systems.
Known Issues
Upgrading Security Suite on systems with OMNIC Specta gives an error message during installation
When upgrading Security Suite on a system that has OMNIC Specta installed and the Thermo Indexer running, you may receive an error message after entering the username and password to use for the Secure Data Storage service.
- Workaround
-
Close Thermo Indexer.
-
Close the error dialog.
-
Press Next > again.
Excluding directories from file monitoring does not work properly
There are multiple limitations when excluding directories from file monitoring in Security Administration:
-
Wildcards in the middle of a directory name are not supported and will not work as expected. For example, excluding
Temp*Dirwill actually exclude all files in any folder containingTempDirin their name, such asMyTempDirFolderorTempDirectory. -
There is an implied leading and trailing asterisk (\*) wildcard for each directory name. For example, excluding
Tempwill actually exclude all files in any folder containingTempin their name, such asMyTempFolderorTemporaryFiles. To only exclude files in a folder named exactlyTemp, you must specify\Temp\(including the backslashes). -
The question mark (?) wildcard is not supported at all. Since the question mark is not a valid character in a directory name on Windows systems, attempting to use it will not exclude any directories.
-
The directory name should not include the drive letter or root backslash. For example, to exclude all files in
C:\Temp, you should only enterTemp\.
Security Suite 2.6 Hotfix 1
-
Build number: 2.6.1.17
-
Release date: July 2024
-
Supported operating systems
-
Server
-
Windows Server 2016 w/ Desktop Experience
-
Windows Server 2019 w/ Desktop Experience
-
Windows Server 2022 w/ Desktop Experience
-
-
Workstation
-
Windows 10 64-bit Version 1607 and newer
-
Windows 11 64-bit
-
-
New Features
None
Resolved Issues
Unable to stop the Security Service while configuring the Audit Log Database
When configuring the Audit Log Database, it could report that it was unable to stop the Security Service. This occurred when using the SQLite database or integrated security with any of the other supported databases.
Known Issues
Upgrading Security Suite on systems with OMNIC Specta gives an error message during installation
When upgrading Security Suite on a system that has OMNIC Specta installed and the Thermo Indexer running, you may receive an error message after entering the username and password to use for the Secure Data Storage service.
- Workaround
-
Close Thermo Indexer.
-
Close the error dialog.
-
Press Next > again.
Unable to add users or groups to Access Control items
When adding users or groups to Access Control items, the user interface would stop responding and never recover.
Erroneous message when trying to save configuration files in OMNIC and OMNIC for Dispersive Raman
When trying to save configuration files in OMNIC and OMNIC for Dispersive Raman, a message would appear stating that the audit log could not be written to, and the operation could not be performed without logging.
The Thermo Scientific Secure Data Storage Service would stop when the Thermo Scientific Security Service was restarted
When the Thermo Scientific Security Service was restarted (e.g. as part of reconfiguring the audit log database), the Thermo Scientific Secure Data Storage Service would stop and not restart automatically.
Excluding directories from file monitoring does not work properly
There are multiple limitations when excluding directories from file monitoring in Security Administration:
-
Wildcards in the middle of a directory name are not supported and will not work as expected. For example, excluding
Temp*Dirwill actually exclude all files in any folder containingTempDirin their name, such asMyTempDirFolderorTempDirectory. -
There is an implied leading and trailing asterisk (\*) wildcard for each directory name. For example, excluding
Tempwill actually exclude all files in any folder containingTempin their name, such asMyTempFolderorTemporaryFiles. To only exclude files in a folder named exactlyTemp, you must specify\Temp\(including the backslashes). -
The question mark (?) wildcard is not supported at all. Since the question mark is not a valid character in a directory name on Windows systems, attempting to use it will not exclude any directories.
-
The directory name should not include the drive letter or root backslash. For example, to exclude all files in
C:\Temp, you should only enterTemp\.
Security Suite v2.6
-
Build number: 2.6.0.78
-
Release Date: June 2024
-
Supported Operating Systems
-
Server
-
Windows Server 2016 w/ Desktop Experience
-
Windows Server 2019 w/ Desktop Experience
-
Windows Server 2022 w/ Desktop Experience
-
-
Workstation
-
Windows 10 64-bit Version 1607 and newer
-
Windows 11 64-bit
-
-
New Features
Added Error Logging to Security Suite
Security Administration, Thermo Scientific Secure Data Storage Service, Thermo Scientific File Change Monitor Service, Thermo Scientific Security Service, and the security client now log generate log files in %ProgramData%\Thermo Scientific\Security Suite to help with troubleshooting issues. The log files have the same name as the executable that generated them and have a .log extension.
Resolved Issues
Improved reliability of audit log entries when network connection is unstable
If the connection to either the audit log service or the database storing audit events was lost after the application started, audit log entries fail to be properly recorded. The security client now retries sending the log entry to the audit log service if a communication error occurs with it or the database. If the entry cannot be sent after a set number of retries, the entry is stored in a cache that will be written once the connection is reestablished.
The event source for some audit events showed the legacy application name
Some audit events showed the legacy application name in the event source field (for example, OMNIC for Dispersive Raman displayed as OMNIC For Nicolet Almega). The event source now shows the same application name as Security Administration for any new entries.
Thermo Scientific File Change Monitor Service could stop before logging all changes
If the Thermo Scientific File Change Monitor Service was stopped while it was processing a large number of changes, it could stop before it finished logging all changes. The service now logs all changes before stopping.
Audit Log connectivity checks could pass when they should fail
If the client was able to communicate with the audit log service, but the service was unable to communicate with the database, the connectivity check would pass. The connectivity check now reports failure for both cases.
Known Issues
Upgrading Security Suite on systems with OMNIC Specta gives an error message during installation
When upgrading Security Suite on a system that has OMNIC Specta installed and the Thermo Indexer running, you may receive an error message after entering the username and password to use for the Secure Data Storage service.
- Workaround
-
Close Thermo Indexer.
-
Close the error dialog.
-
Press Next > again.
Erroneous message when trying to save configuration files in OMNIC and OMNIC for Dispersive Raman
When trying to save configuration files in OMNIC and OMNIC for Dispersive Raman, a message would appear stating that the audit log could not be written to, and the operation could not be performed without logging.
The Thermo Scientific Secure Data Storage Service would stop when the Thermo Scientific Security Service was restarted
When the Thermo Scientific Security Service was restarted (e.g. as part of reconfiguring the audit log database), the Thermo Scientific Secure Data Storage Service would stop and not restart automatically.
Excluding directories from file monitoring does not work properly
There are multiple limitations when excluding directories from file monitoring in Security Administration:
-
Wildcards in the middle of a directory name are not supported and will not work as expected. For example, excluding
Temp*Dirwill actually exclude all files in any folder containingTempDirin their name, such asMyTempDirFolderorTempDirectory. -
There is an implied leading and trailing asterisk (\*) wildcard for each directory name. For example, excluding
Tempwill actually exclude all files in any folder containingTempin their name, such asMyTempFolderorTemporaryFiles. To only exclude files in a folder named exactlyTemp, you must specify\Temp\(including the backslashes). -
The question mark (?) wildcard is not supported at all. Since the question mark is not a valid character in a directory name on Windows systems, attempting to use it will not exclude any directories.
-
The directory name should not include the drive letter or root backslash. For example, to exclude all files in
C:\Temp, you should only enterTemp\.
Security Suite v2.5
-
Build number: 2.5.0.12
-
Release Date: March 2024
-
Supported Operating Systems
-
Server
-
Windows Server 2012 R2
-
Windows Server 2016 w/ Desktop Experience
-
Windows Server 2019 w/ Desktop Experience
-
-
Workstation
-
Windows 10 64-bit Version 1607 and newer
-
Windows 11 64-bit
-
-
New Features
Added Support for New OMNIC Paradigm Features
Added functionality necessary to support new features in OMNIC Paradigm 2.6.
Resolved Issues
None
Known Issues
Upgrading Security Suite on systems with OMNIC Specta gives an error message during installation
When upgrading Security Suite on a system that has OMNIC Specta installed and the Thermo Indexer running, you may receive an error message after entering the username and password to use for the Secure Data Storage service.
- Workaround
-
Close Thermo Indexer.
-
Close the error dialog.
-
Press Next > again.
Excluding directories from file monitoring does not work properly
There are multiple limitations when excluding directories from file monitoring in Security Administration:
-
Wildcards in the middle of a directory name are not supported and will not work as expected. For example, excluding
Temp*Dirwill actually exclude all files in any folder containingTempDirin their name, such asMyTempDirFolderorTempDirectory. -
There is an implied leading and trailing asterisk (\*) wildcard for each directory name. For example, excluding
Tempwill actually exclude all files in any folder containingTempin their name, such asMyTempFolderorTemporaryFiles. To only exclude files in a folder named exactlyTemp, you must specify\Temp\(including the backslashes). -
The question mark (?) wildcard is not supported at all. Since the question mark is not a valid character in a directory name on Windows systems, attempting to use it will not exclude any directories.
-
The directory name should not include the drive letter or root backslash. For example, to exclude all files in
C:\Temp, you should only enterTemp\.
Security Suite v2.4 Hotfix 1
-
Build number: 2.4.1.18
-
Release Date: March 2023
-
Supported Operating Systems
-
Server
-
Windows Server 2012 R2
-
Windows Server 2016 w/ Desktop Experience
-
Windows Server 2019 w/ Desktop Experience
-
-
Workstation
-
Windows 10 64-bit Version 1607 and newer
-
Windows 11 64-bit
-
-
New Features
None
Resolved Issues
Thermo File Change Monitor Service crash on long filenames
The Thermo File Change Monitor Service crashes when it receives a notification of a change to a file where the full path is exactly 260 characters long.
Known Issues
Upgrading Security Suite on systems with OMNIC Specta gives an error message during installation
When upgrading Security Suite on a system that has OMNIC Specta installed and the Thermo Indexer running, you may receive an error message after entering the username and password to use for the Secure Data Storage service.
- Workaround
-
Close Thermo Indexer.
-
Close the error dialog.
-
Press Next > again.
Excluding directories from file monitoring does not work properly
There are multiple limitations when excluding directories from file monitoring in Security Administration:
-
Wildcards in the middle of a directory name are not supported and will not work as expected. For example, excluding
Temp*Dirwill actually exclude all files in any folder containingTempDirin their name, such asMyTempDirFolderorTempDirectory. -
There is an implied leading and trailing asterisk (\*) wildcard for each directory name. For example, excluding
Tempwill actually exclude all files in any folder containingTempin their name, such asMyTempFolderorTemporaryFiles. To only exclude files in a folder named exactlyTemp, you must specify\Temp\(including the backslashes). -
The question mark (?) wildcard is not supported at all. Since the question mark is not a valid character in a directory name on Windows systems, attempting to use it will not exclude any directories.
-
The directory name should not include the drive letter or root backslash. For example, to exclude all files in
C:\Temp, you should only enterTemp\.
Security Suite v2.4
-
Build number: 2.4.0.12
-
Release Date: December 2022
-
Supported Operating Systems
-
Server
-
Windows Server 2012 R2
-
Windows Server 2016 w/ Desktop Experience
-
Windows Server 2019 w/ Desktop Experience
-
-
Workstation
-
Windows 10 64-bit Version 1607 and newer
-
-
New Features
Enabled creation of dump files upon application crash
Set up the system to automatically create dump files in the %ProgramData%\Thermo Scientific\Security Suite\Dump Files folder if any of the applications crash.
Improved error handling
Added top-level error handlers to prevent application crashes from recoverable errors.
Resolved Issues
Missing security policy name on non-English systems
The policy name for "Use exclude list for all events, not just directory renames" was blank in Security Administration when run on non-English systems.
Missing audit log entries for multi-file operations
Certain multi-file operations (copy, move, etc.) could result in some of the affected files being missed in the audit log.
Improperly formed application XML files could cause Security Administrator to crash
An intentionally malicious application definition XML file could cause Security Administrator to crash upon trying to read the file in.
Known Issues
Upgrading Security Suite on systems with OMNIC Specta gives an error message during installation
When upgrading Security Suite on a system that has OMNIC Specta installed and the Thermo Indexer running, you may receive an error message after entering the username and password to use for the Secure Data Storage service.
- Workaround
-
Close Thermo Indexer.
-
Close the error dialog.
-
Press Next > again.
Thermo File Change Monitor Service crash on long filenames
The Thermo File Change Monitor Service crashes when it receives a notification of a change to a file where the full path is exactly 260 characters long.
Excluding directories from file monitoring does not work properly
There are multiple limitations when excluding directories from file monitoring in Security Administration:
-
Wildcards in the middle of a directory name are not supported and will not work as expected. For example, excluding
Temp*Dirwill actually exclude all files in any folder containingTempDirin their name, such asMyTempDirFolderorTempDirectory. -
There is an implied leading and trailing asterisk (\*) wildcard for each directory name. For example, excluding
Tempwill actually exclude all files in any folder containingTempin their name, such asMyTempFolderorTemporaryFiles. To only exclude files in a folder named exactlyTemp, you must specify\Temp\(including the backslashes). -
The question mark (?) wildcard is not supported at all. Since the question mark is not a valid character in a directory name on Windows systems, attempting to use it will not exclude any directories.
-
The directory name should not include the drive letter or root backslash. For example, to exclude all files in
C:\Temp, you should only enterTemp\.
Security Suite v2.3 Hotfix 2
-
Build number: 2.3.2.46
-
Release Date: December 2021
-
Supported Operating Systems
-
Server
-
Windows Server 2012 R2
-
Windows Server 2016 w/ Desktop Experience
-
Windows Server 2019 w/ Desktop Experience
-
-
Workstation
-
Windows 10 64-bit
-
-
New Features
New policy to ignore all file events occurring in any folder from the exclude list.
The Admin program already had a policy that allowed a directory rename events to be suppressed from a list of folders. A new policy has been added that will cause all file events to be suppressed from the folders specified in the exclude list. The policy is under the Admin program and is called "Use exclude list for all events, not just directory renames". Checking the policy will make all folders specified under the "List of directories to exclude from File Change Monitor Service for directory rename events" to be suppressed. By default, this policy is turned off.
Resolved Issues
Renaming file requests with long paths
The Thermo Scientific Secure Data Storage Service could crash if a request was received to rename a file where the source and the destination paths were very long (over 500 characters combined). The buffer size was increased, and checks were added to ensure the combined length of the two paths did not exceed the new limit.
Do not allow services to exit on exception
The Thermo Scientific File Change Monitor Service and the Thermo Scientific Secure Data Storage Service was modified to not exit when an internal exception is encountered. The exceptions are logged, and the services continue.
Record exceptions in unprotected folder
Attempts to log exceptions from the secure file service could fail if the service did not have permission to write to the root of the c: drive. This was fixed by writing the exception log to a folder the service does have permission to write to.
Known Issues
Upgrading Security Suite on systems with OMNIC Specta gives an error message during installation
When upgrading Security Suite on a system that has OMNIC Specta installed and the Thermo Indexer running, you may receive an error message after entering the username and password to use for the Secure Data Storage service.
- Workaround
-
Close Thermo Indexer.
-
Close the error dialog.
-
Press Next > again.
Security Suite v2.3 Hotfix 1
-
Build number: 2.3.1.12
-
Release Date: November 2020
-
Supported Operating Systems
-
Server
-
Windows Server 2012 R2
-
Windows Server 2016 w/ Desktop Experience
-
Windows Server 2019 w/ Desktop Experience
-
-
Workstation
-
Windows 10 64-bit
-
-
New Features
None
Resolved Issues
Rebooting the Security Server halts file system monitoring audit events
When using a separate server for secure data storage, audit events stop being recorded for changes after the security server is restarted until the file server is also restarted.
Using a separate server for secure data storage can fail to connect to the audit log service
The default account used for the file change monitoring service on the file server does not have access rights to the named pipe used by the audit log service.
Known Issues
None
Security Suite v2.3
-
Build number: 2.3.0.10
-
Release Date: July 2020
-
Supported Operating Systems
-
Server
-
Windows Server 2008 R2
-
Windows Server 2012
-
Windows Server 2012 R2
-
Windows Server 2016 w/ Desktop Experience
-
Windows Server 2019 w/ Desktop Experience
-
-
Workstation
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 10 64-bit
-
-
New Features
None
Resolved Issues
Incorrect characters in some event descriptions
When using mixed languages between the application and the server, multi-byte characters may not appear correctly. The malformed characters will not be recoverable.
Known Issues
None
Security Suite v2.2 Hotfix 1
Release Date: Sep-2019
Supported Operating Systems
-
Server
-
Windows Server 2008 R2
-
Windows Server 2012
-
Windows Server 2012 R2
-
Windows Server 2016 w/ Desktop Experience
-
Windows Server 2019 w/ Desktop Experience
-
-
Workstation
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 10 64-bit
-
New Features
None
Resolved Issues
Unable to run ChangeServer.exe
Attempting to run ChangeServer.exe on a Nicolet Summit gave an error message.
Known Issues
None
Security Suite v2.2
Release Date: Jul-2019
Supported Operating Systems
-
Server
-
Windows Server 2008 R2
-
Windows Server 2012
-
Windows Server 2012 R2
-
Windows Server 2016 w/ Desktop Experience
-
Windows Server 2019 w/ Desktop Experience
-
-
Workstation
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 10 64-bit
-
New Features
Added support for OMNIC Paradigm
Added support for OMNIC Paradigm and the new location the security server name can be stored.
Resolved Issues
Updated help file
Updated the online help file with the latest content.
Ensured that all client installations include ChangeServer.exe
Certain installation scenarios would fail to include ChangeServer.exe.
Improved handling of long audit log entries
Audit Log entries with more than 259 characters could not be properly transmitted on to the audit log service.
Known Issues
Unable to run ChangeServer.exe
Attempting to run ChangeServer.exe on a Nicolet Summit gave an error message.
Security Suite v2.1 HF3
Release Date: Sep-2018
Supported Operating Systems
-
Server
-
Windows Server 2008 R2
-
Windows Server 2012
-
Windows Server 2012 R2
-
Windows Server 2016 w/ Desktop Experience
-
-
Workstation
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
-
New Features
None
Resolved Issues
Improved error handling
Improved handling of error conditions and the messages returned to the user.
Known Issues
None
Security Suite v2.1 HF2
Release Date: 11-Apr-2018
Supported Operating Systems
-
Server
-
Windows Server 2008 R2
-
Windows Server 2012
-
Windows Server 2012 R2
-
Windows Server 2016 w/ Desktop Experience
-
-
Workstation
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
-
New Features
None
Resolved Issues
Certain network configurations prohibited retrieving the group membership for an account during installation.
The group membership check is no longer necessary and has been removed to avoid requiring additional permissions during installation.
Known Issues
None
Security Suite v2.1 HF1
Release Date: 28-Feb-2018
Supported Operating Systems
-
Server
-
Windows Server 2008 R2
-
Windows Server 2012
-
Windows Server 2012 R2
-
Windows Server 2016 w/ Desktop Experience
-
-
Workstation
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
-
New Features
None
Resolved Issues
Certain Active Directory security configurations prohibited authenticating the service account during installation.
This was found to be an issue with the authentication options specified when communicating with the domain controller.
Documentation error in setting up database service account
Documentation incorrectly stated that read-only access was required for the service account when read-write permission is correct.
NTFS folder permissions could prevent secure data storage from saving data
The default folder permissions for user folders on newer Windows installations could cause the Secure Data Storage service to be unable to place data files in the secure location.
Known Issues
None
Security Suite v2.1
Release Date: 05-Jan-2018
Supported Operating Systems
-
Server
-
Windows Server 2008 R2
-
Windows Server 2012
-
Windows Server 2012 R2
-
Windows Server 2016 w/ Desktop Experience
-
-
Workstation
-
Windows 7 32-Bit
-
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
-
New Features
Add Audit Manager to view application events
A new application called Audit manager has been added to view events from Thermo Fisher applications. This replaces the use of Windows Event Manager.
Application events now stored in user defined data base
Application events will now be stored in a data base instead of the Windows Event Viewer. The data base is defined at installation.
OQ for OMNIC Security Suite Software has been modified to align with Audit Manager 1.0
The document called OQ for OMNIC Security Suite software has been modified to align with the new Audit Manger tool.
Version 2.0 installation process did not allow the end-user to install instrument software alone
Version 2.1 installation process now allows the end-user to install just the instrument software or Audit Manager.
Resolved Issues
+
Intermittently on a distributed install the file and log service are not set to the account that was specified at install time and are not started.
This was found to be a timing problem and has been resolved for the version 2.1 installation
Known Issues
None
Security Suite v2.0 Hotfix 3
Release Date: 03-Nov-2017
Supported Operating Systems
-
Server
-
Windows Server 2008 R2
-
Windows Server 2012
-
Windows Server 2012 R2
-
Windows Server 2016 w/ Desktop Experience
-
-
Workstation
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
-
New Features
None
Resolved Issues
When installing distributed on some networks a timeout error would occur
When installing a distributed configuration, on some networks when you are at the screen where you enter the name of the computer where Security Administration is installed when you press the Next > button it will not advance and gives the error "Unable to connect - network timeout error". This prevents you from installing the software. The method of verifying the connection has been changed to prevent this problem.
Changes so that secure folder permissions can be altered
Previously when software applications started they altered the folder permissions on the secure folder every time the program started. Now they only alter the permissions the first time when the folder is created or filled with files. This allows IT departments to alter the configuration from the recommended default permissions.
Changes to test secure folder permissions
Now that folder permissions can be altered, the software tests writing to the secure folder using the current account and the specified secure file service account. If the current account is allowed to write to the secure folder a warning message appears stating this is not expected. Also if the secure file service account cannot write to the secure folder an error message occurs as well.
Change to list users/groups on active directory domain with no Organizational Units (OU) defined
Previously if you had an active directory domain that had no organizational units defined an error message "The specified domain does not exist or could not be contacted" popped up when starting Security Administration. After closing the message the software works properly but would not list users and groups on the domain, and required you to type the user or group name. Now the software hides the list of organizational units if none are defined and lists the users and groups when the domain is selected. Normal active directory domains let you choose the domain, followed by the organizational unit, which then lists the users/groups in that organization.
Known Issues
The installer sometimes fails to start the Thermo Scientific File Service and set the services to the account specified at installation time.
When this occurs the Instrument application will display a dialog box stating "Thermo File Service: The service is installed but is not running." If the services are not running and set to the account specified at installation, the instrument application will not be able to save data files to a secure folder.
How to set the File Service and Log Service accounts when they are not set properly
You must be logged in as an administrator on the local machine and you must know the service account name and password to proceed.
-
In the "Search programs and files" edit box on Win7 or the Win10 Cortana "ask me anything" box type "services" and hit return. This brings up the services program.
-
Select a service and press the letter "T" to find the "Thermo Scientific File Service"
-
Right click on the "Thermo Scientific File Service" and "Thermo Scientific Log Service" and choose "Stop" to stop both services.
-
Now right click on the Thermo Scientific File service and choose "Properties". Select the "Log On" tab and select the "This account" radio button. Type in the name of the service account that will be granted read-write access to the secure data storage directory. Enter in the password and confirm the password. Press Ok.
-
Right click on the Thermo Scientific Log service and choose "Properties". Select the "Log On" tab and select the "This account" radio button. Type in the name of the service account that will be granted read-write access to the secure data storage directory. Enter in the password and confirm the password. Press Ok.
-
Right click on the Thermo Scientific File Service and choose "Start",
-
Finally, right click on the Thermo Scientific Log Service and choose "Start".
This procedure only needs to be done once after installation if the services are not configured properly at installation. This occurs intermittently but has been observed.
Security Suite v2.0 Hotfix 2
Release Date: 31-Aug-2017
Supported Operating Systems
-
Server
-
Windows Server 2008 R2
-
Windows Server 2012
-
Windows Server 2012 R2
-
Windows Server 2016 w/ Desktop Experience
-
-
Workstation
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
-
New Features
None
Resolved Issues
Under some circumstances, the Save button does not save changes
Doing the following sequence in Security Administration will not save the changes:
-
Open one of the Access Control items.
-
Change the permission from Deny to Allow or Allow to Deny.
-
Click on the Save button.
Thermo Log Service could generate extraneous file events in the log
In certain circumstances, the Thermo Log Service would generate extraneous file events in the log when a file was being saved by an application.
Unable to install newer software after setting up Secure Data Folders
If the default data directory was chosen as the Secure Data Folder, the user would be unable to install newer versions of OMNIC, qPro, INSIGHT, etc. The user is now prohibited from selecting one of these well-known directories as the Secure Data Folder.
The installation could get stuck if the user went back to the account selection screen
During distributed installations, going back to the service account selection screen would leave the user unable to go forward again and would state that the username or password was incorrect every time.
Known Issues
None
Security Suite v2.0 Hotfix 1
Release Date: 20-June-2017
Supported Operating Systems
-
Server
-
Windows Server 2008 R2
-
Windows Server 2012
-
Windows Server 2012 R2
-
Windows Server 2016 w/ Desktop Experience
-
-
Workstation
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
-
New Features
None
Resolved Issues
Previous security settings not retained during upgrade
Upgrading to Security Suite 2.0 would not properly retain the previous settings.
Automatic load of applications
The automatic loading of applications into Security Administration during installation on non-English locales would not load any application that did not have a configuration file for the current locale.
Known Issues
Under some circumstances, the Save button does not save changes
Doing the following sequence in Security Administration will not save the changes:
-
Open one of the Access Control items.
-
Change the permission from Deny to Allow or Allow to Deny.
-
Click on the Save button.
- Workaround
-
Selecting File → Save from the menu or click on any other object before selecting the Save button.
Security Suite v2.0
Release Date: 14-June-2017
Supported Operating Systems
-
Server
-
Windows Server 2008 R2
-
Windows Server 2012
-
Windows Server 2012 R2
-
Windows Server 2016 w/ Desktop Experience
-
-
Workstation
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
-
New Features
Added support for Active Directory
After selection of a domain, it tests if the domain supports Active Directory. If it does, a dropdown is displayed with Organizational Units. This allows you to list just the users and groups associated with an Organizational Unit instead of an entire domain.
Improve ease of use with access rights settings
Changed the access rights selection to have a dropdown with 2 choices "Allow" or "Deny" to clarify the use of access rights. Previously it used a checkbox for the allow/deny choice.
Audit event for program entry and exit
Add audit events when logging on or exiting Security Administration.
Access to audit events
Added menu item to automatically call up the Windows event viewer to view the audit events for Thermo Fisher products.
Automatically load applications on first install
Changes to automatically load the Application XML files after installation when you run Security Administration after installation. This simplifies setup and insures that new capabilities are merged into existing installations.
Change directory renames
Change the Thermo Log Service to have directory rename events be a different event type to allow filtering these items out.
Allow ignore specific directories
Add a new policy which lets you edit a list of directories that the Thermo Log Service ignores. This list will allow the asterisk to be used as a wildcard character and will default to the directories already rejected.
Require full setup
Security Administration will not allow you to close until all secure directories have been defined for applications that support secure directories.
Simplify User/Group changes to access control items
Add a new menu item that allows you to replace a user/group with another user/group for all access control items. This allows built-in groups like "Administrators" to be easily replaced with a custom group appropriate for your network.
Simplify common secure folder setup
Changed to set a common folder location for all subfolders for application data using an environment variable and show that path after it is set. When the instrument software is started for the first time, it will create this common secure folder location, copy the files from the local installation (C: drive) to the secure location, and finally it will set the correct permissions for that folder. Afterwards, the folders will be set to read only for all users except for the one account specified at installation time to have read/write permission.
Make security server name more obvious
Display the name of the Security server so customers know what to type when running the ChangeServer program or installing the instrument software.
Resolved Issues
Google temporary directories in log
Improved the logging service to ignore renames of Google chrome temporary directories.
Server name dialog sometimes inaccessible
Changed the ChangeServer program so that the dialog for specifying the server name is a topmost window that cannot go behind another window.
Warn event log overwrite
Display an error message if the event log is set to overwrite old events rather than archive them. Also provide a menu and toolbar item to perform configuration tests.
Improved user interface
Made various controls wider and taller to allow more text to be displayed without being cut off.
Improved printing
Improved the printing of application settings so that items are not cut off and when printing data folders it, displays the root path separately from the folder path.
Date print out format
Fixed issue where the date format was not displayed properly in printouts if the date format is configured to use text names for months and not numbers.
Event Logging
Changed event logging to always use the security server to ensure that events generated by non-admin users are recorded properly.
Known Issues
None
Security Administration v1.8
Release Date: Jan-2017
Supported Operating Systems
-
Server
-
Windows Server 2008 R2
-
Windows Server 2012
-
Windows Server 2012 R2
-
-
Workstation
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
-
New Features
Enhance for use with INSIGHT
Changed the ThermoFileService.exe and ThermoClient.dll to support the INSIGHT software using secure folders to store data on networks.
Resolved Issues
Rebuilt all binaries so that they have matching 1.8 version numbers
Previously most of the files were version 1.6 and only the file watcher was version 1.7, this was confusing for some customers and has been resolved.
Security Administration program displays an error message on start
On some Win 10 systems an error message stating "The API return buffer is too small" appears on some domains. After you close the error message the software appears to operate properly. This has been resolved.
The Thermo File Service did not always start upon reboot of the computer
The changes to the file watcher process sometimes prevented the service from starting when the computer was rebooted. This has been resolved.
Known Issues
None
Security Administration v1.7
Release Date: Jul-2016
Supported Operating Systems
-
Windows XP SP3 32-bit
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
New Features
Enhance for use with OMNICxi map files
OMNICxi map files, *.mapx, are now supported in the file watcher process.
Resolved Issues
None
Known Issues
None
Security Administration v1.6 Hotfix 2
Release Date: Oct-2011
Supported Operating Systems
-
Windows XP SP3 32-bit
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
New Features
Enhance for use with OMNICxi map files
OMNICxi map files, *.mapx, are now supported in the file watcher process.
Resolved Issues
The installer for Security Administration did not uninstall previous versions before it installed the current version
The installer for the software now properly uninstalls the previous version before it installs the current version. The software being installed has not changed.
Known Issues
None
Security Administration v1.6 Hotfix 1
Release Date: Oct-2011
Supported Operating Systems
-
Windows XP SP3 32-bit
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
New Features
None
Resolved Issues
Logon attempts now match the Windows Security policy
Unsuccessful login attempts now match that of the Windows Security policy.
This only occurs in this version
Known Issues
Policy items configured to be accessible only to members of the Windows built-in Administrators group may not be accessible to users who are members of that group
|
Note
|
This issue only occurs on Windows Vista and newer. It does not occur on Windows XP. |
When using Security Administration to configure policy items, if you configure a policy item to be accessible only to members of the Windows built-in Administrators group, the policy item may not be accessible to users who are members of that group.
This happens because starting in Windows Vista administrators no longer start with administrator privileges.
For those who encounter this problem, there are two workarounds:
-
Explicitly start OMNIC (or other Thermo Security controlled software) using the right-click "Run as Administrator" option.
-
Revise the "Advanced Properties" of the shortcut on the desktop for the Thermo Security controlled software by setting the checkmark for the "Run as Administrator" option.
Using either of these workarounds the software will behave as it did in Windows XP, and the software will start with administrative privileges.
Security Administration 1.6
Resolved Issues
Changed occurrences of Thermo Electron to Thermo Scientific
Changed the splash screen, icon, and about box to display Thermo Scientific. The about box now displays information about the installed version, not the file version.
Display error message when adding optional software before main software
The administration application did not add application information properly if you added an optional software package before you added the package it depended upon. Now if you attempt to add an optional feature for an application before you add the main application you will receive the message, "You must add the (application name) application before you can add this file to security administration."
Setup failed to install over existing installations
Security Administration 1.5 did not install over previous installations because the installer said, "A later version of Security Administration is already installed on this machine." The installer on the CD was not the correct version. This has been changed.
Known Issues
None.
Security Administration 1.5
Resolved Issues
Unable to change access rights for some groups on German Windows
When using the German version of Windows and the computers are connected to a domain network, the built-in groups (such as Users and Administrators) cannot be added or removed from the access rights list. This problem only occurred on the German version of Windows when networked. The 1.5 release fixes this issue.
New Features
Support for Windows Vista Business software.
The Security Administration software has been changed to fix issues noted during operation using the Vista operating system. This includes changing the help system from .HLP files to .CHM files.
Known Issues
Events in event log have server name for the computer name on Vista operating system
If you install Security Administration on a computer with the Windows Vista operating system and are using a network, audit events are different from in previous versions. In this circumstance, audit events will correctly have the name of the remote user of the computer that generated the event, but will list the name of the Security Administration computer rather than the name of the remote computer. This change is required because Windows Vista does not allow remote computers to write to the event log.
Adding software options before the software they reference does not work
When adding applications to Security Administration, if you add an optional software package, such as OMNIC for Raman or OMNIC Series, before you add the OMNIC application, the system policies will not be installed.
Security Administration 1.4
Resolved Issues
Guest user accounts were allowed access
If guest user accounts were enabled (they are disabled by default and it is recommended that they remain disabled), those accounts could have access rights even though Guest accounts were denied access. This issue was caused by a mistake in the default XML files for OMNIC software products. However, the 1.4 version of Security Administration corrects XML files that have grant and deny items in the wrong order.
New Features
Support for TEVA software.
The Security Administration software has added the ability to store application-specific passwords as policy items. Several new types of audit events for the TEVA software were also added.
Support for Chinese and Russian.
Chinese and Russian translations of Security Administration have been added to the CD, bringing the number of translated languages to seven.
Known Issues
There are no known issues in this release.
Security Administration 1.3
Resolved Issues
Cannot add a domain account to a policy group
Under some conditions, after a local user account has been added to a policy group, attempts to add a domain user account will fail. This has been fixed.
Cannot add "Power Users" group to policies or access rights
If you add the "Power Users" group to Access rights or policy groups when you save, the "Power Users" group will not be saved.
Cannot load multiple languages of the same application XML file
If you load a translation of an application XML file after already loading a different language, multiple entries will occur for the same items. Now you can load the English XML file of an application, followed by a foreign language, and it will switch the display to that foreign language without changing any settings. You can also load the English XML file after this and it will switch back to English.
Adding new signature reasons doesn’t allow access by default
If you add a new signature reason, it does not show up by default in the application. The default for new signature reasons was no access rights; you needed to add users or groups who could access the signature reason. In 1.3 this behavior was changed to add the "Users" group to a new signature reason as the default. If you do not wish to allow all users to access the signature reason, simply delete the "Users" group.
Security Administration allows any valid user account to log in
When the Security Administration program starts, it prompts for a user name and password. However, it accepts any valid user name and password account on the Administration computer rather than just the name and password for the currently logged-on user. This has been changed so that only the person currently logged on to the computer can log into Security Administration.
New Features
Support for RESULT software.
The Security Administration software has added the ability to add several new types of policy items, such as items from a drop-down list. Several new types of audit events for RESULT software were also added. The Security Administration program has a new menu item to remove applications from administration.
Security Administration 1.2a
Resolved Issues
Japanese characters do not display properly in user and group list
Some Japanese characters are not displayed properly in the list of available users and groups. The Microsoft default list only partially supports Japanese characters and needed a fix to work with the full character set.
New Features
This release has the same feature set as 1.2. There are no new features in this release.
Security Administration 1.2
Resolved Issues
Administrator program requires user name and password at startup.
The Security Administration program now requires user authentication when the program starts to verify the user identity of the Security Administration software.
Administrator program handles large networks better.
The Security Administration program will switch to initiating the accounts on the local computer, rather than the accounts on the login domain if the enumeration takes a long time. An edit control has been added that allows user names to be entered manually so enumeration can be avoided. Accounts are not re-enumerated when switching from the access control portion of the tree to the policy portion. These changes make using the software faster on large global networks that take minutes to enumerate the available users and groups.
Administrator program easily adds or removes access control entries for all access control items.
The Security Administration program has two new menu items: The Add to All Access Control Items command will add an Allow or Deny entry for the specified user to every function in the Access Control section of the current application. The Remove From All Access Control Items command will remove the entry for a specified user or group from all functions in the Access Control section of the current application.
Administrator program recognizes more foreign language countries.
The Security Administration program now supports choices such as French (Canadian) instead of only the direct foreign language choices of French, German, etc.
Improved consistency in printing to different printers
Text would print much larger on some inkjet printers than it did on laser printers. Now correctly calculates font sizes.
New Features
Support for VISION software.
Some capabilities were added to get the date/time of the server.
Security Administration 1.1
Resolved Issues
User’s full name not always retrieved
In some circumstances the full name of the user account was not retrieved and the logon name was used instead. Now it properly gets the full name of the account if specified for use in audit events.
New Features
Added support for several foreign languages
Added support for foreign language translations.
How to Contact Us
Current contact information is located at https://www.thermofisher.com
Select the "Contact Us" icon at the top of the screen
Attachment(s):
| File | Last Modified |
|---|---|
| Security Suite Release Notes.pdf | July 29, 2026 |