Thermo Fisher Scientific
AN 50875 - The Use of FT-Raman Spectroscopy and Chemometric Procedures in the Analysis of Pharmaceuticals
Description: 
The FT-Raman technique has many benefits in the analyticallaboratory, including speed, easy sample preparation andspectral information that is interpreted in the same way asFT-IR spectra