The Use of FT-Raman Spectroscopy in the Study of Formulated Pharmaceuticals
Description:
This note examines the advantages that the Fourier transform Raman (FT-Raman) technique offers pharmaceutical studies. It discusses the collection of spectra from formulated pharmaceuticals and draws comparisons between FT-Raman and Fourier transformation infrared (FT-IR) absorption measurements.
Environment:
FT-Raman
Attachment(s):
File | Last Modified |
---|---|
The Use of FT-Raman Spectroscopy in the Study of Formulated Pharmaceuticals.pdf | July 28, 2022 |