IR2323 - DXR Microscope Shipping Restraint
Description:
The RIM (Raman Injection Module) mirror flipper assembly can become permanently damaged if the DXR Microscope is transported without first inserting the shipping restraint plug.
If you have misplaced the shipping restraint, a new one (470-391100) can be ordered.
The stage and microscope nosepiece must also be removed before transporting.
Environment:
DXR Raman Microscopes
Attachment(s):
File | Last Modified |
---|---|
IR2323 - DXR Microscope Shipping Restraint.pdf | June 12, 2022 |