IR2345 - DXR Light Leaks
Description:
Due to the high sensitivity of the updated CCD on the DXR Raman spectrometers, some customers in high ambient light conditions have seen light affect their spectra. The updated CCD camera is installed on DXR system serial numbers > AIY1000500 and > AIZ1000500. Older systems that have received the spectrograph update (spectrograph serial numbers > AJA100500) will also need the gasket kit installed.