Running Wavelength Performance Verification of using a Holmium Glass filter
Description:
Follow these steps to run wavelength performance verification of the GENESYS 10S using a Holmium glass filter:
- Press the TEST key.
- Use the down arrow key to highlight Performance Verification (Enter).
- Use the down arrow key to highlight Wavelength Accuracy Standards (Enter).
- Press Delete nm 3 times to delete the 3 old peaks.
- Press Add nm 638.0 (Enter).
- Press 1 (Enter).
- Press Add nm 445.8 (Enter).
- Press 1 (Enter).
- Press Add nm 279.4 (Enter).
- Press 1 (Enter).
- Verify the measurement mode is set to Absorbance Peak.
- In the sample compartment leave the B position empty (no cuvette).
- Place the Holmium Oxide filter in the 1 position.
- Press the Start Test key.
Following the execution of this procedure, the instrument will run all three peaks and give you Pass/Fail results.
Environment:
GENESYS 10S