Thermo Fisher Scientific
Performance Verification Kit
Description: 
These are the contents of part number 840-312600, GENESYS 40-180 Performance Verification Kit:
840-312600 GENESYS 40-180 Performance Verification Kit
Use with the built in performance verification tests in the Diagnostics method of the GENESYS 40, 50, 140, 150, 180 or BioMate 160 to perform fundamental PV tests to verify correct instrument operation.
Includes:
  • 3 neutral density filters calibrated for absorbance at 440, 465, 546.1, 590 and 635 nm
  • Nominal ND filter values: 0.5A, 1.0A, 2.0A
  • Calibration traceable through NIST via ISO/IEC Standard 17025 accredited process
  • Didymium glass filter for wavelength accuracy determination
  • Wavelength standard calibration traceable through primary standard mercury lamp emission peaks via ISO/IEC Standard 17025 accredited process.
  • 3 Stray Radiant Energy Filters at 220, 340, and 400 nm, not calibrated or traceable
Environment:
  • GENESYS 40, 50, 140, 150, 180
  • BioMate 160