Color calculations
Description:
To perform color calculations, scan samples using INSIGHT software with the proper scan range for the color calculation you are making. Save the scan as a CSV file. After collecting scan, open the ColorCalc software and load the CSV file. The spectrum can now be manipulated with ColorCalc and various color systems can be used to make the final calculations. See ColorCalc Help menu.
Environment:
- Evolution 200 Series: Evolution 201, Evolution 220, Evolution 260
- VISIONlite ColorCalc software with a revision number below 5.2
Attachment(s):
File | Last Modified |
---|---|
INSIGHT Export Scan data.pdf | December 27, 2021 |