Thermo Fisher Scientific
Blank display at power up
Issue
The display remains blank, and the instrument will not initialize. 
Environment
  • GENESYS 40-180
  • BioMate 160
Resolution
  • Turn the instrument off.
  • Install the paper into the internal printer.
  • Turn the instrument on.
Cause
If the internal printer is installed in the instrument and there is no paper in the printer when the instrument is turned on, the display remains blank, and the instrument will not initialize.