Thermo Fisher Scientific
Error 1330
Issue
Error 1330 when installing Insight
Environment
PC has Windows 7 or 10, 32-bit Starter
Resolution
Upgrade the PC operating system to Windows 7 or 10, 64-bit Professional
  • Was this article helpful?