Evolution 220, 260 Beam Dimensions
Issue
What are the beam dimensions of the Evolution 220 and Evolution 260 instruments?
Environment
- Evolution 220
- Evolution 260
Resolution
SETTING | SLIT SIZE | BEAM PATCH AT SAMPLE |
1 nm | 0.158 mm x 3 mm | 0.46 x 2.5 mm |
2 nm | 0.340 mm x 3 mm | 0.6 x 2.5 mm |
Microcell | 0.340 mm x 1.6 mm | 0.6 x 1.96 mm |
Fiber (round) | 0.88 mm round | 1.5 x 1.8 mm |
Materials | 3 mm round | 2.8 x 2 mm |