Thermo Fisher Scientific
Failed wavelength accuracy when using a didymium glass standard
Issue
The GENESYS 40 and 140 fail wavelength accuracy when using a didymium glass standard.
Environment
  • GENESYS 40
  • GENESYS 140
Resolution
Use the Lambda interference filter from the Spec Standards 2 kit to test wavelength in the G40 and G140. 
  • Was this article helpful?