GENESYS and BioMate Beam Dimensions
Issue
What are the beam dimensions of the GENESYS and BioMate instruments?
Environment
- GENESYS 40
- GENESYS 50
- GENESYS 140
- GENESYS 150
- GENESYS 180
- BioMate 160
Resolution
MEASUREMENTS TAKEN AT 550 nm | MEASUREMENTS TAKEN USING A FOCUSED BEAM | ||
INSTRUMENTS | Z Height | Beam Height | Beam Width |
GENESYS 40 | 8.5 mm | 4.37 mm | 1.99 mm |
GENESYS 140 | 8.5 mm | 4.37 mm | 1.99 mm |
GENESYS 150 | 8.5 mm | 4.13 mm | 2.23 mm |
GENESYS 180 | 8.5 mm | 4.13 mm | 2.23 mm |
BioMate 160 | 8.5 mm | 4.13 mm | 2.23 mm |