Thermo Fisher Scientific
XE Calibration button grayed out
Issue
Why is the XE Calibration button grayed out when Insight is connected to an Evolution 350?
Environment
  • Evolution 350
  • Insight v2.4.174
Resolution
  • The Xe lamp is not used to calibrate the wavelength for the Evolution 350.
  • The XE Calibration button is supposed to be grayed out, this is normal.
  • Use a Hg lamp to calibrate the wavelength on an Evolution 350. 
  • Was this article helpful?