Modifying ascapp.ini file to automatically export Quant sample results
Issue
How can sample result files be automatically exported?
Environment
- GENESYS 10S
- GENESYS 20
- GENESYS 30
- Evolution 60S
- Evolution 300
- Evolution 600
Resolution
The default settings for VISIONlite is to make available the option to export sample results into a CSV file in Fixed and Scan modes.
Quant mode does allow the use of the “Export results” function in the File menu. However, the option is only active directly after a measurement.
By modifying the ascapp.ini file, this option can be activated to automatically export sample result CSV files.
Refer to the attached document for instructions on modifying the ascapp.ini file.
Attachment(s)
| File | Last Modified |
|---|---|
| Modifying ascapp.ini file to automatically export Quant sample results.pdf | December 25, 2021 |