CAD OQ - Noise Test Failing
Issue
Trouble getting Noise test to pass for CAD qualification.
Environment
- Qualification (OQ)
- Corona Veo Charged Aerosol Detectors (CAD)
- Vanquish CAD
Resolution
Use better water for the test.
A highly pure water such as:
Fisher Plasma Water (1L): 080157
Agilent Ultra Pure Water for HPCE (500mL): THRAG-5062-857
Details: https://www.agilent.com/store/undefi...8578/5062-8578
Can be ordered through Cloud 4 Service if the customer doesn't have suitable water on hand.
Analysis
Using a higher quality of water can help achieve significantly better Noise and Drift values. Most laboratories do not have pure enough water for testing.
Also, always make sure the instrument has had a proper PM (if not new), and confirm no environmental factors (HVAC, for example) are not affecting the unit's operation as these can impact the drift test as well.
Cause
CAD instruments are highly sensitive to the quality of water, and require a higher purity than most LC detectors.
Attachment(s)
File | Last Modified |
---|---|
Plasma Water.png | September 28, 2022 |
Agilent THRAG-5062-8578.png | July 18, 2022 |