Add-on Drivers Required for Surveyor FL Plus and Surveyor RI Plus
Description:
Support for the Surveyor FL Plus and the Surveyor RI Plus detectors is not included in the base version of ChromQuest 5.0. The add-on versions for ChromQuest 4.2 that are included with the detectors do not work on ChromQuest 5.0.
Eventually the Surveyor FL Plus Add-on for CQ 5.0 will be CHROM-98012 and the Surveyor RI Plus Add-on for CQ 5.0 will be CHROM-98014. They will ship with the new units when they are available.
Environment:
- ChromQuest 5.0
- Surveyor FL Plus
- Surveyor RI Plus
Attachment(s):
File | Last Modified |
---|---|
0001453 FLpluS_Installer_HotFix_CQ50.zip | January 20, 2022 |
0001453 RIPlusInstall_5.zip | January 20, 2022 |