065448 Part A - Rev 7.12 - Chromeleon 2013 Thermo Fisher Scientific Operational Qualification and Performance Qualification Software Templates for ICS-3000 Instruments - User's Guide
The Thermo Fisher Scientific ICS-3000 System OQ/PQ User’s Guide provides detailed instructions for conducting Operational Qualification (OQ) and Performance Qualification (PQ) on ICS-3000 instruments. It includes test procedures to verify system performance, e.g., flow rate accuracy, eluent generator current, baseline noise and drift, injector precision, sample carryover, and detector linearity. The manual outlines the preparation of standards and eluents, system setup, and troubleshooting.
Description:
This manual introduces the conventions used in performing OQ/PQ with the ICS-3000 systems; including the basic requirements, industry standards and manual summary. For the ICS-3000 system, electrochemistry cells in Integrated Amperometry mode are supported for qualification tests. For the Integrated Amperometry detection qualification a
pH-Ag-AgCl reference electrode is required for all tests. The ICS-3000 electrochemistry detector DC amperometry and cyclic voltammetry modes are not supported for OQ-PQ testing.
Environment:
- ICS-3000 systems
- Chromeleon 7