065448 Part A - Rev 7.30 - Chromeleon Operational Qualification and Performance Qualification Software Templates for ICS-3000 Instruments - User's Guide
Step-by-step guidance covers preparing Chromeleon and ICS-3000 hardware and generating qualification sequences with the Instrument Qualification Wizard, then run OQ/PQ tests in the prescribed order. It details module compatibility, required kits, sample layouts, and pass/fail limits. It also shows how to use the Instrument Qualification Wizard to auto-build sequences and run them in order, with troubleshooting and example reports for GLP/cGMP records.
Description:
The purpose of PQ is to prove and document that an analytical system functions according to a specification that is suitable for the system’s routine operation. A system is subject to wear when being operated. PQ should be performed at regular intervals after the initial installation and OQ. Thermo Fisher Scientific recommends performing PQ every six months.
Environment:
- Chromeleon 7.1
Module Minimum Chromeleon Software Version
- AS-AP 7.1 SR1 DU0C
- AS 7.1 SR1
- AS-DV 7.1 SR1
- PDA-100 7.1 SR1
- ICS-3000 SP or DP 7.1 SR1
- ICS-3000 EG 7.1 SR1
- ICS-3000 DC 7.1 SR1
- ICS-3000 CD 7.1 SR1
- ICS-3000 ED 7.1 SR1
- FLUKE189/289 7.1 SR1
- ICS Series VWD 7.1 SR1
- ICS Series PDA 7.1 SR1
- ICS-3000 TC 7.1 SR1
- RF-2000 with UCI-50, UCI-100 or UI20 interface 7.1 SR1
- Pump:
- ICS-3000 SP
- ICS-3000 DP
- Manual injection or any of the following autosamplers:
- AS
- AS-DV
- AS-AP
- AS40
- AS-AP autosampler:
- 10 mL tray PN 074938 with ICS-3000 CD pull or push mode
- 1.5 mL tray PN 074936 with ICS-3000 CD and ED push mode only - The optional conductivity pH module for the AS-AP is currently not supported in OQ-PQ.
- Absorbance detector:
- ICS Series VWD
- ICS Series PDA
- Conductivity detector: ICS-3000 CD
- Electrochemical detector: ICS-3000 ED
- Fluorescence detector: RF-2000 with
- UCI-50 interface
- UCI-100 interface
- UI-20 interface
- Eluent Generator: ICS-3000 EG
Attachment(s):
| File | Last Modified |
|---|---|
| 065448 Part A - Rev 7.30 - Chromeleon Operational Qualification and Performance Qualification Software Templates for ICS-3000 Instruments - User's Guide.PDF | January 06, 2022 |