Thermo Fisher Scientific
Agilent Drivers for Chromeleon 2.2 Update 1
Description: 
The following Agilent components are included in Agilent Drivers for Chromeleon 7 (Rev. 2.2 Update 1):
  • Agilent Instrument Control Framework 2.6 Update 1 
  • Agilent LC Drivers A.02.19 SR2 
  • Agilent ELSD Drivers A.01.07 DU1 
  • Agilent GC-HS Drivers 3.0 SR4
Environment:
  • Agilent ICF
  • Chromeleon 7
  • Was this article helpful?