Thermo Fisher Scientific
Agilent Drivers for Chromeleon 2.3
Description: 
A new set of drivers to control Agilent LC and GC hardware. New features include:
  • Dual Simultaneous Injection for GCs
  • Dynamic Licensing (see Change Detail below)
  • Adding lines to running sequences
  • Separate user guides for GC and LC
Please see the Release Notes provided with the drivers for more new features and further details on these features.
Environment:
  • Chromeleon 7.2 SR5 (including all MUs)
  • Chromeleon 7.2.10 (including all MUs)
  • Chromeleon 7.3
  • Was this article helpful?