4828.3250A - Rev 9.1 - Thermo Scientific Chromeleon for HPLC - Release Notes
Use Chromeleon 7’s Instrument Qualification Wizard to create and run OQ/PQ sequences. Prepare pumps, autosamplers, detectors, and column ovens, configure the system, and follow supplied templates. Observe module-specific steps and limitations, especially for Vanquish ovens and Agilent ICF. Execute tests, review reports, and repeat failed checks. Avoid editing ICF methods, and use only supported version combinations.
Description:
Release Notes 9.1
Environment:
Thermo Scientific Chromeleon HPLC
Attachment(s):
| File | Last Modified |
|---|---|
| 4828.3250A - Rev 9.1 - Thermo Scientific Chromeleon for HPLC - Release Notes.PDF | January 24, 2022 |