Agilent Drivers for Chromeleon 1.2
This package supplies drivers that allow Chromeleon 7 to control Agilent chromatography instruments via the Agilent instrument control framework. It provides a list of supported instruments and firmware versions, along with detailed documentation and step-by-step installation guidance. A readme file offers remarks and updates. The distribution also contains software licensing terms for review and compliance. Review all guides before install to confirm set up compatibility with confidence.
Description:
The following Agilent components are included in Agilent Drivers for Chromeleon 7 (Rev. 1.2):
- Agilent Instrument Control Framework A.02.05
- Agilent LC Drivers A.02.18
- Agilent ELSD Drivers 1.7
- Agilent LC Modules/Firmware (see “Supported Agilent LC Modules and Firmware” on page 10)
Environment:
- Agilent ICF
- Chromeleon 7
Attachment(s):
| File | Last Modified |
|---|---|
| Agilent-Drivers-1.2.zip | January 19, 2022 |