Thermo Fisher Scientific
Cat File for Chinese Language Chromeleon
Description: 
为以下 Chromeleon 中文版提供 cat 文件。这样,用户就可以在 Thermo Scientific 247 IC 上安装任意这些版本
A cat file is available for the following Chinese Language Chromeleon Versions. This enables the user to install any of those versions on a Thermo Scientific 247 IC
Chromeleon 7.2.10 MUa
Chromeleon 7.2.10 MUd
Chromeleon 7.2.10 MUf
Chromeleon 7.2.10 MUg
Chromeleon 7.3.0
Chromeleon 7.3.0 MUb
Chromeleon 7.3.1
Chromeleon 7.3.2
所有后续的中文版都已包含该 cat 文件,因此可以直接安装
All later Chinese Versions will already have the cat file included and can thus be installed without further issue
Environment:
中文版 Chromeleon 7 
  • Was this article helpful?