Chromeleon 7.3.2 MUf Hotfix 939441 Software and Release Notes
Description:
Chromeleon 7.3.2 MUf Hotfix 939441 Software and Release Notes
When attempting to set folders to read-only as part of a workflow, it was observed that folders containing signed sequences could not be set to read-only from a remote client. Instead, the following error message was displayed:
"The read-only operation cannot be performed at this time because some of the selected sequences have been signed."
Environment:
Chromeleon 7.3.2 MUf
Attachment(s):
| File | Last Modified |
|---|---|
| Release Notes - Hotfix 939441 for Chromeleon 7.3.2 MUf.pdf | October 16, 2026 |
| Hotfix 939441 for Chromeleon 7.3.2 MUf.zip | October 16, 2026 |