Chromeleon Registry Keys for TNG Tune
TNG Tune will only be accessible from within Chromeleon for compliance purposes.
Description:
A registry key determines whether TNG Tune will open inside a Console tab or as it's own window outside of Chromeleon. If the following key exists, Tune will open as it's own window:
Computer\HKEY_LOCAL_MACHINE\SOFTWARE\WOW6432Node\Thermo\SII Xcalibur
Environment:
- Chromeleon SII
- Xcalibur
- TSQ Systems
Attachment(s):
| File | Last Modified |
|---|---|
| clipboard_e7a9dd82eb436340fdf971d2cb8ae49bd.png | November 07, 2025 |
| SII for Xcalibur, Chromeleon Registry Keys for TSQ Tune.pdf | November 14, 2021 |