Thermo Fisher Scientific
TraceFinder 4.1 Release Notes
Description: 
These release notes briefly list features in the Thermo TraceFinder 4.1 data system and known issues that still exist in the TraceFinder application.
Environment:
TraceFinder
Hardware:
  • 2.33 GHz dual-core processor enabled
  • 8 GB RAM with system managed memory enabled
  • DVD drive
  • Resolution display 1280 ×1024 SXGA
  • 5 GB available on drive C
  • NTFS format
Software:
  • Microsoft Windows 7 SP1 Professional (64-bit) English only
  • Microsoft .NET Framework 4.5.1
  • Adobe™ Reader™ 10.1
  • Thermo Scientific software:
    • Xcalibur 4.0
    • Foundation platform 3.1 SP3
    • mzVault 1.0 SP1
    • FreeStyle 1.1 SP1
    • LC Devices 3.1 or later
    • Exactive Series 2.7or later
    • TSQ Endura TSQ Quantiva 2.0 or later
    • TSQ 2.6 or later
    • Fusion 1.2 SP1 or later
    • Aria MX 2.2 or later
    • GC Devices 3.2 or later
    • ISQ 3.2 SP1 or later
    • SII 1.2 for Xcalibur or later
    • TSQ 8000 3.2 or later
    • LTQ Series 2.7 SP4 or later
  • Was this article helpful?