XCALI-97991 - Rev A - Xcalibur 4.2 Release Notes
Description:
These release notes briefly list changes in the Thermo Xcalibur data system version 4.2, fixes to previously reported issues, and known issues still existing in the software.
Environment:
Xcalibur 4.2
Hardware:
- 3.6 GHz Quad core processor with 16 GB RAM
- Video card and monitor capable of 1920 ×1080 resolution
- 1 TB hard drive
- CD/R-ROM drive or DVD
Software:
- Microsoft Windows 7 Professional SP1 64-bit or Windows 10 Enterprise 2016 LTSB
- .NET Framework 4.6.2
- Thermo Foundation 3.1 SP5 or later
- Microsoft Office 2013
- Adobe Acrobat or Reader 10.1
Attachment(s):
File | Last Modified |
---|---|
XCALI-97991 - Rev A - Xcalibur 4.2 Release Notes.PDF | December 29, 2021 |